Skip to Content
Merck
All Photos(1)

Key Documents

68250

Sigma-Aldrich

Methyl Orange

for microscopy (Hist.), indicator (pH 3.0-4.4)

Synonym(s):

4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid sodium salt, Acid Orange 52, Helianthin, Orange III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N3NaO3S
CAS Number:
Molecular Weight:
327.33
Colour Index Number:
13025
Beilstein:
4732884
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02
Pricing and availability is not currently available.

grade

for microscopy (Hist.)
indicator (pH 3.0-4.4)

Quality Level

form

powder

technique(s)

titration: suitable

pH

3.0-4.4, pink to yellow

solubility

methanol: water (1:1): 0.1 g/10 mL, orange to very deep orange

λmax

501.4 nm in H2O (acidified)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)S([O-])(=O)=O

InChI

1S/C14H15N3O3S.Na/c1-17(2)13-7-3-11(4-8-13)15-16-12-5-9-14(10-6-12)21(18,19)20;/h3-10H,1-2H3,(H,18,19,20);/q;+1/p-1/b16-15+;

InChI key

STZCRXQWRGQSJD-GEEYTBSJSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Methyl orange is an orange colored synthetic dye,[1] which undergoes a change in color depending on the pH of the solution.[2] It basically develops a red color in acidic solution and yellow color in basic solution.[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Longman Science Chemistry 10 (2008)
Science for Tenth Class Part 2 Chemistry null
Lin Lin et al.
Nanoscale, 5(2), 588-593 (2012-12-04)
Shell@core-nanostructured TiO(2)@ZnO n-p-n heterojunction nanorods with diameter of 30 nm were successfully fabricated via a hydrothermal method. The photodegradation rate of the TiO(2)@ZnO n-p-n nanorods evaluated by photodegrading methyl orange has been demonstrated to increase three times compared to that
Ya Yang et al.
Nano letters, 13(2), 803-808 (2013-01-18)
In general, methyl orange (MO) can be degraded by an electrocatalytic oxidation process driven by a power source due to the generation of superoxidative hydroxyl radical on the anode. Here, we report a hybrid energy cell that is used for
Jie Li et al.
Nanotechnology, 24(15), 155603-155603 (2013-03-23)
We report on the synthesis of high-quality microporous/mesoporous BN material via a facile two-step approach. An extremely high surface area of 1687 m(2) g(-1) and a large pore volume of 0.99 cm(3) g(-1) have been observed in the synthesized BN porous whiskers. The formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service