Skip to Content
Merck
All Photos(1)

Documents

E-002

Supelco

Ecgonine methyl ester-D3 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Ecgonine methyl ester-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14D3NO3
CAS Number:
Molecular Weight:
202.27
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Ecgonine methyl ester is a major urinary metabolite of cocaine, a popular illicit drug well known as "crack" or the free base form of cocaine that can be smoked. This stable-labeled internal standard is suitable for use in cocaine testing or isotope dilution methods for GC/MS or LC/MS applications in urine drug testing, clinical toxicology or forensic analysis.

Application


  • Refinement of cocaine metabolite analysis: Ecgonine methyl ester-D₃ is used as a deuterated internal standard to spike urine, plasma, or serum in a method developed for the quantitation of cocaine and its metabolites in urine and blood using gas chromatography-mass spectrometry (GC-MS (Fleming et al., 2010).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service