Skip to Content
Merck
All Photos(1)

Key Documents

B-007

Supelco

Benzoylecgonine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO4
CAS Number:
Molecular Weight:
289.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3ccccc3)C(O)=O

InChI

1S/C16H19NO4/c1-17-11-7-8-12(17)14(15(18)19)13(9-11)21-16(20)10-5-3-2-4-6-10/h2-6,11-14H,7-9H2,1H3,(H,18,19)/t11-,12+,13-,14+/m0/s1

InChI key

GVGYEFKIHJTNQZ-RFQIPJPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoylecgonine is the primary metabolite of cocaine and a major biomarker for determination of cocaine use. This certified reference standard is suitable for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing. Benzoylecgonine is also the main pharmaceutical ingredient in the prescription drug Esterom, a topical analgesic used for the relief of muscle pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata Bystrowska et al.
Pharmacological reports : PR, 64(6), 1337-1349 (2013-02-15)
We employed a cocaine intravenous self-administration model based on positive reinforcement of animals' instrumental reactions (i.e., lever pressing) rewarded by a dose of the drug. We also carried out simultaneous characterization of the pharmacokinetics of cocaine and its metabolites in
Robert L Fitzgerald et al.
Journal of analytical toxicology, 36(2), 106-111 (2012-02-18)
Our objective was to develop a "dilute and shoot" liquid chromatography-tandem mass spectrometry confirmatory procedure that uses full scan product ion spectra to identify drugs that are present above cutoff values as determined by isotope dilution relative to a deuterium-labeled
Lubertus Bijlsma et al.
The Science of the total environment, 443, 200-208 (2012-11-29)
In this work, ultra-high-performance liquid chromatography (UHPLC) coupled to a hybrid quadrupole time-of-flight mass spectrometer (QTOF MS) has allowed the discovery and elucidation of degradation products of cocaine and its main metabolite benzoylecgonine (BE) in water. Spiked surface water was
Sara Castiglioni et al.
Environmental science & technology, 47(3), 1452-1460 (2012-12-25)
The aim of this study was to integrally address the uncertainty associated with all the steps used to estimate community drug consumption through the chemical analysis of sewage biomarkers of illicit drugs. Uncertainty has been evaluated for sampling, chemical analysis
Juliet Kinyua et al.
Journal of forensic sciences, 57(5), 1349-1353 (2012-04-19)
Indirect estimation methods of cocaine consumption may not reflect the real extent of cocaine use. Another approach is sewage epidemiology. This direct approach is based on analysis of a stable cocaine metabolite, benzoylecgonine (BE), in wastewater. Influent to the Lubbock

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service