Skip to Content
Merck
All Photos(1)

Key Documents

E-001

Supelco

Ecgonine methyl ester solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Ecgonine methyl ester solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

N1(C2[C@H]([C@H](CC1CC2)O)C(=O)OC)C

InChI

1S/C10H17NO3/c1-11-6-3-4-7(11)9(8(12)5-6)10(13)14-2/h6-9,12H,3-5H2,1-2H3/t6?,7?,8-,9+/m0/s1

InChI key

QIQNNBXHAYSQRY-ABIFROTESA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution standard suitable for use in LC/MS or GC/MS testing applications including forensic analysis, urine drug testing, and clinical toxicology. Ecgonine methyl ester is a major urinary metabolite of cocaine, a popular illicit drug well known as "crack" or the free base form of cocaine that can be smoked.

Application


  • Ecgonine methyl ester standard:The site- and species-specific hydrolysis rates of cocaine, demonstrating the importance of Ecgonine methyl ester as a reference standard in understanding cocaine metabolism pathways, aiding in pharmaceutical and forensic applications (Szocs et al., 2017).

  • Mass spectrometry reagent:Sensitive fluorescence probe was developed using molecularly imprinted polymers and Mn-doped ZnS quantum dots for the determination of cocaine and metabolites in urine, highlighting the use of Ecgonine methyl ester as a crucial mass spectrometry reagent for drug testing (Chantada-Vazquez et al., 2016).

  • Cocaine metabolite analysis:Magnetic molecularly imprinted polymers were developed for solid phase extraction of cocaine and metabolites in urine before high-performance liquid chromatography - tandem mass spectrometry, illustrating the essential role of Ecgonine methyl ester in the selective detection and quantification of cocaine metabolites (J Sanchez-Gonzzlez, et al., 2016).

  • Biochemical research chemicals:Porous membrane-protected molecularly imprinted polymer micro-solid-phase extraction method for analyzing urinary cocaine and its metabolites, validating Ecgonine methyl ester′s utility in biochemical and pharmacological studies (Sanchez-Gonzalez et al., 2015).

  • Forensic toxicology supplies:Employed peptides to trap cocaine, using solid phase extraction followed by liquid chromatography-mass spectrometry in plasma samples, underlining the application of Ecgonine methyl ester in forensic toxicology for precise and accurate drug monitoring (Mascini et al., 2013).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marta Concheiro et al.
Journal of analytical toxicology, 31(9), 573-580 (2007-12-21)
A method, using 0.5 mL of urine, was developed for the simultaneous determination of ecgonine methyl ester, benzoylecgonine, morphine, codeine, 6-acetylmorphine, amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxymethamphetamine (MDMA), methadone, 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP), and d-lysergic acid diethylamide (LSD). The analysis was performed by
Cláudia Margalho et al.
Journal of forensic and legal medicine, 18(3), 132-135 (2011-03-23)
Postmortem tissues (e.g. liver, kidney) have been long used in forensic applications especially in those cases where blood is unavailable. The aim of this paper is to demonstrate the importance of the information provided to the forensic toxicologist at the
Rosa Cordero et al.
Journal of analytical toxicology, 34(9), 543-548 (2010-11-16)
The concentrations and ratios of cocaine, benzoylecgonine (BE), and ecgoninemethylester (EME) in 360 head hair segments and 34 pubic hair samples collected at coroner's postmortem examinations were reviewed. The cases included diverse histories and causes/circumstances of death. The hair was
C Pistos et al.
Rapid communications in mass spectrometry : RCM, 23(23), 3772-3780 (2009-11-11)
At present, no official criteria exist for drug identification using single quadrupole mass spectrometers although the European Union (EU) criteria for compound identification have been adopted. These criteria are evaluated with respect to the confirmation of cocaine and its metabolites
Philippe Hantson et al.
Journal of forensic and legal medicine, 18(8), 385-387 (2011-10-25)
Life-threatening complications may occur in body-packers and the rupture of a single packet containing cocaine may lead to fatality. We report the case of a 35-year-old body-packer who developed at the airport clinical signs of cocaine toxicity. There was evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service