Skip to Content
Merck
All Photos(1)

Documents

B-004

Supelco

Benzoylecgonine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO4
CAS Number:
Molecular Weight:
289.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CN1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3ccccc3)C(O)=O

InChI

1S/C16H19NO4/c1-17-11-7-8-12(17)14(15(18)19)13(9-11)21-16(20)10-5-3-2-4-6-10/h2-6,11-14H,7-9H2,1H3,(H,18,19)/t11-,12+,13-,14+/m0/s1

InChI key

GVGYEFKIHJTNQZ-RFQIPJPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference standard is suitable for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Benzoylecgonine is the primary metabolite of cocaine and a major biomarker for the determination of cocaine use. Benzoylecgonine is also the main pharmaceutical ingredient in the prescription drug Esterom, a topical analgesic used for the relief of muscle pain.

Application

The certified reference solution (CRM) in solution can also be used as follows:
  • Development and validation of a high-performance liquid chromatography-diode array detector (HPLC-DAD) based method to determine cocaine, its two main degradation products, and the common adulterants found in cocaine-based products from 115 seized samples
  • Analysis of benzoylecgonine in human urine samples using a monolithic molecularly imprinted polymer (MIP) for the solid phase microextraction (SPME) followed by UV detection
  • Separation and determination of cocaine and its metabolites― benzoylecgonine, ecgonine methyl ester, and cocaethylene in hair samples by hydrophilic interaction liquid chromatography-tandem mass spectrometry (HILIC-MS/MS)
  • Estimation of cocaine, benzoylecgonine, cocaethylene, ecgonine methyl ester (EME), and norcocaine in dried blood spots (DBS) by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Multi-residue analysis of xylazine, free morphine, 6-acetylmorphine, codeine, cocaine, and benzoylecgonine in post-mortem blood samples by ultra-high-pressure liquid chromatography-electrospray ionization (UPLC-ESI) and triple-quadrupole tandem mass spectrometry

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Bouvarel et al.
Talanta, 233, 122611-122611 (2021-07-04)
A simple, selective, and sensitive method involving a miniaturized solid phase extraction step based on a monolithic molecularly imprinted polymer (MIP) directly coupled on-line to UV detection was developed for the determination of benzoylecgonine (BZE) in complex biological samples. Monolithic
N Nikolettos et al.
Clinical and experimental obstetrics & gynecology, 39(1), 36-42 (2012-06-09)
To investigate the response of pregnant and non pregnant rat myometrium to benzoylecgonine, a cocaine metabolite, and oxytocin and to investigate the efficiency of ritodrine and atosiban to overcome the effects of benzoylecgonine and oxytocin. Isolation of rat myometrial tissue
Beata Bystrowska et al.
Pharmacological reports : PR, 64(6), 1337-1349 (2013-02-15)
We employed a cocaine intravenous self-administration model based on positive reinforcement of animals' instrumental reactions (i.e., lever pressing) rewarded by a dose of the drug. We also carried out simultaneous characterization of the pharmacokinetics of cocaine and its metabolites in
Charles A Dackis et al.
Journal of substance abuse treatment, 43(3), 303-312 (2012-03-02)
This is a randomized, double-blind, placebo-controlled study of modafinil treatment for cocaine dependence. Patients (N = 210) who were actively using cocaine at baseline were randomized to 8 weeks of modafinil (0 mg/day, 200 mg/day, or 400 mg/day) combined with
Sara Castiglioni et al.
Environmental science & technology, 47(3), 1452-1460 (2012-12-25)
The aim of this study was to integrally address the uncertainty associated with all the steps used to estimate community drug consumption through the chemical analysis of sewage biomarkers of illicit drugs. Uncertainty has been evaluated for sampling, chemical analysis

Articles

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service