Skip to Content
Merck
All Photos(1)

Documents

A-009

Supelco

6-Acetylmorphine, 1.0 mg/mL solution

1 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

6-Acetylmorphine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4OC(C)=O)c35

InChI

1S/C19H21NO4/c1-10(21)23-15-6-4-12-13-9-11-3-5-14(22)17-16(11)19(12,18(15)24-17)7-8-20(13)2/h3-6,12-13,15,18,22H,7-9H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1

InChI key

JJGYGPZNTOPXGV-SSTWWWIQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Acetylmorphine, also known as 6-monoacetylmorphine or 6-MAM, is an active metabolite of heroin and biomarker for heroin usage. This Certified Spiking Solution® is suitable for use as a starting material in calibrators and controls for GC/MS or LC/MS methods in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring. It is relatively difficult to detect in urine samples as it has a short detection window. Within minutes after heroin administration (intravenously), 6-acetylmorphine appears in the saliva and is found at a higher concentration than in plasma.

Application


  • Forensic toxicology reference: 6-Acetylmorphine is used as a key analytical reference material in forensic toxicology to identify heroin use in morphine-positive urine specimens, aiding in the differentiation between pharmaceutical and illicit opioid consumption (Vikingsson et al., 2024).

  • Opiate research chemical: This compound serves as a high-purity standard for opioid research, facilitating the study of opioid metabolism and the development of new analytical techniques in bioanalytical studies (Liu et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTIONS is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High prevalence of 6-acetylmorphine in morphine-positive oral fluid specimens.
Presley
Forensic Science International, 133(1-2) (2003)
Forensic drug testing for opiates: I. Detection of 6-acetylmorphine in urine as an indicator of recent heroin exposure; drug and assay considerations and detection times.
Cone EJ
Journal of Analytical Toxicology (1991)
Joseph Jones
Journal of Analytical Toxicology, 171-175 (2002)
Forensic Science International null
Cai Tie et al.
Journal of mass spectrometry : JMS, 47(11), 1429-1434 (2012-11-14)
With the combination of high separation ability of capillary electrophoresis (CE) and strong identification ability of mass spectrometry (MS), CE/MS is becoming a powerful tool for polar and ionic analytes analysis. Different interfaces have been developed to enhance the sensitivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service