Skip to Content
Merck
All Photos(1)

Key Documents

632090

Sigma-Aldrich

1,4-Diethynylbenzene

96%

Synonym(s):

1,4-Bis(ethynyl)benzene, 1,4-Diacetylenebenzene, 1,4-Diacetylenylbenzene, 1,4-Diethynylbenzene, p-Diethynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4C≡CH
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

94-98 °C (lit.)

storage temp.

2-8°C

SMILES string

C#Cc1ccc(cc1)C#C

InChI

1S/C10H6/c1-3-9-5-7-10(4-2)8-6-9/h1-2,5-8H

InChI key

MVLGANVFCMOJHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Advanced Sensor Development: A novel approach to creating sensitive and selective copper(II) sensors on carbon fiber microelectrodes using 1,4-Diethynylbenzene. This research enhances the performance and application range of electrochemical sensors, particularly in environmental monitoring and industrial processes (Nudurupati et al., 2023).
  • High-Performance OLED Materials: Synthesis of highly phosphorescent dimers of PtAu(2) complexes using 1,4-Diethynylbenzene. This research opens new pathways for creating more efficient and durable organic light-emitting diodes (OLEDs), which are crucial for the development of next-generation display and lighting technologies (Zheng et al., 2022).
  • Mixed-Valence Metal Complexes: A comprehensive study on the electronic structure and mixed-valence characteristics of 1,4-diethynylbenzene-bridged bimetallic complexes. This research provides insights into the tuning of electronic properties for potential applications in molecular electronics and catalysis (Safari et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tan L H Doan et al.
Chemistry, an Asian journal, 10(12), 2660-2668 (2015-08-11)
New Zr(IV)- and Hf(IV)-based metal-organic framework photocatalysts, termed VNU-1 and VNU-2 (where VNU = Vietnam National University), were synthesized and their resulting structures fully characterized. By employing a highly π-conjugated linker, namely 1,4-bis(2-[4-carboxyphenyl]ethynyl)benzene, the optical absorption properties were effectively red-shifted
Shasha Wang et al.
Scientific reports, 7(1), 12712-12712 (2017-10-07)
We developed an efficient one-pot metal-free click polymerization procedure for the synthesis of 3,5-disubstituted polypyrazoles with high yields, high molecular weights, and narrow molecular weight distribution. The method involved two click reactions in a one-pot synthesis. The first reaction was
Mingming Guan et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(6), 625-630 (2019-02-05)
A novel "on-off-on" super-sensitive conjugated polymer fluorescence sensor (PPE-DPA) was developed and it was applied to realize the continuous recognition of Cu2+ and pyrophosphate (PPi). The fluorescence intensity decreased linearly with the change of Cu2+ from 0.05 to 5.0 μmol
Hongyu Mi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 176, 168-173 (2017-01-17)
In this work, a turn on fluorescent sensor, based on Hg
José V Prata et al.
Journal of fluorescence, 30(1), 41-50 (2019-12-08)
A new conjugated polymer possessing calix[4]arene-oxacyclophane units wired-in-series by phenyleneethynylene linkers was synthesized by a Sonogashira-Hagihara cross-coupling method in high yield. The polymer was structurally characterized by FTIR and 1H/13C/HSQC NMR techniques, and its average Mn (38.5 kDa) retrieved from GPC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service