Skip to Content
Merck
All Photos(5)

Key Documents

208671

Sigma-Aldrich

Bis(triphenylphosphine)palladium(II) dichloride

98%

Synonym(s):

Dichlorobis(triphenylphosphine)palladium(II), Palladium(II)bis(triphenylphosphine) dichloride, PdCl2(PPh3)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2PdCl2
CAS Number:
Molecular Weight:
701.90
Beilstein:
4935975
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

Cl[Pd]Cl.c1(P(c2ccccc2)c3ccccc3)ccccc1.c4(P(c5ccccc5)c6ccccc6)ccccc4

InChI

1S/2C18H15P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

YNHIGQDRGKUECZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Bis(triphenylphosphine)palladium(II) dichloride ((PdCl2(PPh3)2) is an organopalladium complex containing tertiary phosphine ligands that is commonly used in C-C bond forming reactions such as Heck arylation, Suzuki coupling, Stille coupling, and Sonogashira coupling.[1]

Application

Bis(triphenylphosphine)palladium(II) dichloride can be used as a catalyst in the:
  • Coupling of 2-iodoanisole and terminal alkynes to synthesize 2,3-disubstituted benzofurans.[2]
  • Copper-free Sonogashira cross-coupling reaction to synthesize diphenylacetylene.[3]
  • Regioselective hydrocarboxylation of styrene.[4]
  • Negishi coupling of fluoroarylzinc pivalates to prepare fluorinated oligophenyls.[5]
  • Coupling of iodo-α-β-unsaturated esters to afford tetrasubstituted olefins.[6]
For small scale and high throughput uses, product is also available as ChemBeads (927759)

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenoxide-assisted P-C bond cleavage in PdCl2 (PPh3) 2 under very mild conditions
Yasuda H, et al.
Journal of Organometallic Chemistry, 691, 1307-1310 (2006)
Modular Synthesis of Phosphino Hydrazones and Their Use as Ligands in a Palladium-Catalysed Cu-Free Sonogashira Cross-Coupling Reaction
Baweja S, et al.
ChemPlusChem, e202300163-e202300163 (2023)
Synthesis of 2, 3-Disubstituted Benzofurans by the Palladium-Catalyzed Coupling of 2-Iodoanisoles and Terminal Alkynes, Followed by Electrophilic Cyclization: 3-Iodo-2-phenylbenzofuran
Yao T, et al.
J. Org. Synth., 91, 283-292 (2014)
PdCl2 (PPh3)2-heteropolyacids-catalyzed regioselective hydrocarboxylation of styrene
J. Mol. Catal. A: Chem., 182, 195-207 (2002)
Synthesizing Highly Fluorinated Oligophenyls via Negishi Coupling of Fluoroarylzinc Pivalates
Engelage E
Synthesis, 54, 711-722 (2021)

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is Product 208671, Dichlorobis(triphenylphosphine)palladium(II), cis or trans?

    1 answer
    1. We have not specifically tested the material in regards to what isomer it is, however, based on the synthesis of it, it is believed to be predominately trans.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. What is Product 208671, Bis(triphenylphosphine)palladium(II) dichloride soluble in?

    1 answer
    1. It is soluble in chloroform, toluene, and benzene.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service