Recommended Products
Assay
98%
mp
53-56 °C (lit.)
SMILES string
OCC(CO)c1ccccc1
InChI
1S/C9H12O2/c10-6-9(7-11)8-4-2-1-3-5-8/h1-5,9-11H,6-7H2
InChI key
BPBDZXFJDMJLIB-UHFFFAOYSA-N
Application
2-Phenyl-1,3-propanediol may be used to synthesize enantiomers of 2-phenyl-3-hydroxypropylcarbamate, via a chemoenzymatic method. It may also be employed in the preparation of 2-phenyl-1,3-propanedithiol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis, characterization and electrochemistry of phenyl-functionalized diiron propanedithiolate complexes.
Polyhedron, 67, 416-421 (2014)
Veterinarni medicina, 36(2), 65-69 (1991-02-01)
In practical farming conditions of an agricultural enterprise situated in South-East Moravia, the repellents N,N'-diethyl-m-toluamide and 2-phenyl propanediol 1,3 were tested after their application to grazing first-calves of the Bohemian Pied breed. The experiment was conducted in July and August
A chemoenzymatic synthesis of both enantiomers of 2-phenyl-3-hydroxypropylcarbamate, a metabolite of felbamate.
Tetrahedron Asymmetry, 6(7), 1765-1772 (1995)
Bioscience, biotechnology, and biochemistry, 71(8), 1858-1864 (2007-08-11)
Bacillus cereus 809A and Burkholderia sp. 711C were isolated from soil. These strains demonstrate hydrolysis activity towards prochiral 2-phenyl-1,3-propanediol diacetate and accumulated the corresponding chiral monoacetates into the reaction mixture. When 2-phenyl 1,3-propanediol diacetate was used as a substrate, the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service