Skip to Content
Merck
All Photos(1)

Key Documents

414670

Sigma-Aldrich

1-Iodo-2-(trimethylsilyl)acetylene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CI
CAS Number:
Molecular Weight:
224.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

130 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

functional group

iodo

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#CI

InChI

1S/C5H9ISi/c1-7(2,3)5-4-6/h1-3H3

InChI key

HNIRHTRSZDSMOF-UHFFFAOYSA-N

Application

1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elodie E Marlier et al.
Inorganic chemistry, 51(4), 2079-2085 (2012-02-11)
A model for cobalamin was synthesized using a new monoanionic tetradentate nitrogen donor ligand; 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine (Tol-BDI((2-pp)2)H) (1), which utilizes isopropylpyridines as pendant arms on a β-diketiminate (BDI) backbone. During the synthesis of 1, the rearrangement product, Tol-BDI((2-pp)(4-pp))H (2) was observed.
Novel polyacetylenes containing pendant 1-pyrenyl groups: synthesis, characterization, and thermal and optical properties.
Rivera E, et al.
Polymer, 43(19), 5059-5068 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service