Skip to Content
Merck
All Photos(1)

Documents

244481

Sigma-Aldrich

1-(Trimethylsilyl)propyne

99%

Synonym(s):

Trimethyl-1-propynylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C≡CSi(CH3)3
CAS Number:
Molecular Weight:
112.24
Beilstein:
1071311
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

99%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

99-100 °C (lit.)

density

0.758 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC#C[Si](C)(C)C

InChI

1S/C6H12Si/c1-5-6-7(2,3)4/h1-4H3

InChI key

DCGLONGLPGISNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(Trimethylsilyl)propyne was used in the synthesis of highly substituted indenes via palladium-catalyzed carboannulation and indenones via a rhodium-catalyzed reaction with 2-bromophenylboronic acids.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyuki Harada et al.
Journal of the American Chemical Society, 129(17), 5766-5771 (2007-04-10)
The Rh-catalyzed reaction of alkynes with 2-bromophenylboronic acids involves carbonylative cyclization to give indenones. The key steps in the reaction involve the addition of an arylrhodium(I) species to an alkyne and the oxidative addition of C-Br bonds on the adjacent
Tetrahedron Letters, 33, 5969-5969 (1992)
Journal of the American Chemical Society, 105, 7473-7473 (1983)
Journal of Polymer Science Part A: Polymer Chemistry, 25, 1353-1353 (1987)
Daohua Zhang et al.
The Journal of organic chemistry, 72(1), 251-262 (2006-12-30)
The synthesis of highly substituted indenes has been achieved by three different transition metal-mediated methods. The first method involves the palladium-catalyzed carboannulation of internal alkynes. The second method utilizes a two-step approach, which involves first the palladium/copper-catalyzed cross-coupling of terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service