Skip to Content
Merck
All Photos(2)

Key Documents

208264

Sigma-Aldrich

Allyltrimethylsilane

98%

Synonym(s):

3-(Trimethylsilyl)propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2Si(CH3)3
CAS Number:
Molecular Weight:
114.26
Beilstein:
906755
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

84-88 °C (lit.)

density

0.719 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)CC=C

InChI

1S/C6H14Si/c1-5-6-7(2,3)4/h5H,1,6H2,2-4H3

InChI key

HYWCXWRMUZYRPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyltrimethylsilane is a general reagent to introduce allyl groups across acid chlorides, aldehydes, ketones, iminium ions, enones, and for cross-coupling with other carbon electrophiles. It is used as a reagent in Hosomi−Sakurai reaction.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general and efficient FeCl3-catalyzed nucleophilic substitution of propargylic alcohols
Zhan Z, et al.
The Journal of Organic Chemistry, 71(21), 8298-8301 (2006)
Ming Yu et al.
Organic letters, 5(24), 4639-4640 (2003-11-25)
[reaction: see text] Direct allylation of glycal-derived donor-acceptor cyclopropanes has been achieved with TiCl(4) activation followed by addition of allyltrimethylsilane. The alpha diastereomer is the major product, with selectivities ranging from 3:1 to 10:1 and yields around 80%.
Ionic liquid-promoted, highly regioselective heck arylation of electron-rich olefins by aryl halides
Mo J, et al.
Journal of the American Chemical Society, 127(2), 751-760 (2005)
Roman Schowner et al.
Angewandte Chemie (International ed. in English), 59(2), 951-958 (2019-11-28)
The origin of hydroxyl group tolerance in neutral and especially cationic molybdenum imido alkylidene N-heterocyclic carbene (NHC) complexes has been investigated. A wide range of catalysts was prepared and tested. Most cationic complexes can be handled in air without difficulty
Control of alpha/beta stereoselectivity in Lewis acid promoted C-glycosidations using a controlling anomeric effect based on the conformational restriction strategy.
Satoru Tamura et al.
Angewandte Chemie (International ed. in English), 42(9), 1021-1023 (2003-03-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service