Skip to Content
Merck
All Photos(1)

Key Documents

L106

Sigma-Aldrich

Loxapine succinate salt

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClN3O · C4H6O4
CAS Number:
Molecular Weight:
445.90
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: soluble

originator

Watson

SMILES string

CN(CC1)CCN1C2=NC3=C(C=CC=C3)OC4=C2C=C(Cl)C=C4.OC(CCC(O)=O)=O

InChI

1S/C18H18ClN3O.C4H6O4/c1-21-8-10-22(11-9-21)18-14-12-13(19)6-7-16(14)23-17-5-3-2-4-15(17)20-18;5-3(6)1-2-4(7)8/h2-7,12H,8-11H2,1H3;1-2H2,(H,5,6)(H,7,8)

InChI key

YQZBAXDVDZTKEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D2/D4 dopamine receptor antagonist; 5-HT2A/2B, 5-H7 serotonin receptor antagonist; dibenzoxazepine antipsychotic agent.

Features and Benefits

This compound was developed by Watson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert H Howland
Journal of psychosocial nursing and mental health services, 50(1), 16-18 (2012-01-11)
Acute agitation in patients with schizophrenia or bipolar disorder is an important clinical management problem. Liquid concentrates, orally disintegrating tablets, and/or intramuscular formulations of several second-generation atypical antipsychotic drugs are available for treating acute agitation. Loxapine is an older first-generation
[Progress through formulation].
Heike Oberpichler-Schwenk
Medizinische Monatsschrift fur Pharmazeuten, 36(7), 243-243 (2013-08-01)
B L Roth et al.
Psychopharmacology, 120(3), 365-368 (1995-08-01)
The affinities of 13 atypical and 12 typical antipsychotic drugs for the cloned rat D4 dopamine receptor and the D4/D2 ratios were examined. Of the atypical antipsychotic drugs tested, only clozapine, risperidone, olanzapine, zotepine and tiospirone had affinities less than
B Biton et al.
The Journal of pharmacology and experimental therapeutics, 340(3), 706-715 (2011-12-16)
Sodium-activated potassium (K(Na)) channels have been suggested to set the resting potential, to modulate slow after-hyperpolarizations, and to control bursting behavior or spike frequency adaptation (Trends Neurosci 28:422-428, 2005). One of the genes that encodes K(Na) channels is called Slack
F-R Cousin et al.
L'Encephale, 38(1), 64-74 (2012-03-03)
Data concerning the clinical and therapeutic characteristics of patients with schizophrenia treated by antipsychotic in naturalistic conditions are useful. Two national pharmacoepidemiological studies were conducted in France, a retrospective survey RÉALITÉ and a prospective study RÉALITÉ LT, to examine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service