Skip to Content
Merck
All Photos(3)

Key Documents

P1793

Sigma-Aldrich

Pimozide

≥98% (TLC), powder, D₂ dopamine receptor antagonist

Synonym(s):

1-[1-[4,4-bis(4-Fluorophenyl)butyl]-4-piperidinyl]-1,3-dihydro-2H-benzimidazol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29F2N3O
CAS Number:
Molecular Weight:
461.55
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Pimozide,

form

powder

solubility

DMSO: 18 mg/mL(lit.)
chloroform: 5 mg/mL
H2O: insoluble(lit.)

originator

Teva

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(CCCN2CCC(CC2)N3C(=O)Nc4ccccc34)c5ccc(F)cc5

InChI

1S/C28H29F2N3O/c29-22-11-7-20(8-12-22)25(21-9-13-23(30)14-10-21)4-3-17-32-18-15-24(16-19-32)33-27-6-2-1-5-26(27)31-28(33)34/h1-2,5-14,24-25H,3-4,15-19H2,(H,31,34)

InChI key

YVUQSNJEYSNKRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D2 dopamine receptor antagonist; binds with high affinity to the cloned 5-HT7 receptor; Ca2+ channel antagonist; antipsychotic

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Cyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Teva. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holger H H Erb et al.
PloS one, 15(8), e0237248-e0237248 (2020-08-14)
Despite enzalutamide's efficacy in delaying the progression of metastatic castration-resistant prostate cancer (CRPC), resistance to this anti-androgen inevitably occurs. Several studies have revealed that the signal transducer and activator of transcription (STAT) 5 plays a role in tumour progression and
Junjun Chen et al.
Chemistry & biology, 18(11), 1390-1400 (2011-11-29)
Ubiquitin-specific proteases (USPs) have in recent years emerged as a promising therapeutic target class. We identified selective small-molecule inhibitors against a deubiquitinase complex, the human USP1/UAF1, through quantitative high throughput screening (qHTS) of a collection of bioactive molecules. The top
L A Opler et al.
The Journal of clinical psychiatry, 52(5), 221-233 (1991-05-01)
Pimozide, a diphenylbutylpiperidine neuroleptic which is FDA-approved as a backup treatment for Gilles de la Tourette's syndrome, has been used abroad for many years as a treatment of schizophrenia and has been recently reported to be particularly effective in treating
Avril Coghlan et al.
PLoS neglected tropical diseases, 17(9), e0011205-e0011205 (2023-09-05)
Hundreds of millions of people worldwide are infected with the whipworm Trichuris trichiura. Novel treatments are urgently needed as current drugs, such as albendazole, have relatively low efficacy. We have investigated whether drugs approved for other human diseases could be
Erik A Nelson et al.
Blood, 117(12), 3421-3429 (2011-01-15)
The transcription factor STAT5 is an essential mediator of the pathogenesis of chronic myelogenous leukemia (CML). In CML, the BCR/ABL fusion kinase causes the constitutive activation of STAT5, thereby driving the expression of genes promoting survival. BCR/ABL kinase inhibitors have

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service