K2628
Ketotifen fumarate salt
≥99% (TLC), powder, H₁ histamine receptor antagonist
About This Item
Recommended Products
Product Name
Ketotifen fumarate salt,
form
powder
Quality Level
originator
Novartis
SMILES string
OC(=O)\C=C\C(O)=O.CN1CCC(\CC1)=C2/c3ccccc3CC(=O)c4sccc24
InChI
1S/C19H19NOS.C4H4O4/c1-20-9-6-13(7-10-20)18-15-5-3-2-4-14(15)12-17(21)19-16(18)8-11-22-19;5-3(6)1-2-4(7)8/h2-5,8,11H,6-7,9-10,12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+
InChI key
YNQQEYBLVYAWNX-WLHGVMLRSA-N
Gene Information
human ... HRH1(3269) , PDE7B(27115)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Biochem/physiol Actions
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service