Skip to Content
Merck
All Photos(1)

Key Documents

31622

Supelco

Bithionol

VETRANAL®, analytical standard

Synonym(s):

2,2′-Thio-bis(4,6-dichlorophenol), Bis(2-hydroxy-3,5-dichlorophenyl) sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl4O2S
CAS Number:
Molecular Weight:
356.05
Beilstein:
2003535
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

parasites

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

enzyme | inhibits
protein synthesis | interferes

SMILES string

Oc1c(Cl)cc(Cl)cc1Sc2cc(Cl)cc(Cl)c2O

InChI

1S/C12H6Cl4O2S/c13-5-1-7(15)11(17)9(3-5)19-10-4-6(14)2-8(16)12(10)18/h1-4,17-18H

InChI key

JFIOVJDNOJYLKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bithionol is a dichlorophenol compound that shows antimicrobial property.1
Chemical structure: diphenylsulfide

Application

Bithionol has been used as reference standard for measuring the concentration of antihelmintic substance in pharmacy products using HPLC.2
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Yang et al.
Neuropharmacology, 51(4), 896-906 (2006-08-01)
The Slack (Sequence like a calcium-activated K channel) (Slo2.2) gene is abundantly expressed in the mammalian brain and encodes a sodium-activated K+ (KNa) channel. Although the specific roles of Slack channel subunits in neurons remain to be identified, they may
Ok-Jae Lee et al.
Journal of gastroenterology and hepatology, 21(10), 1631-1633 (2006-08-25)
Fascioliasis is not common in humans and, furthermore, its ectopic migration into the pancreas is extremely rare. A definitive diagnosis of ectopic fascioliasis is based on the demonstration of flukes in the affected organ. If the flukes invade the parenchyma
R L Florent et al.
Journal of fish diseases, 32(5), 391-400 (2009-02-27)
This study examined the efficacy of bithionol as a prophylactic or therapeutic oral treatment for Atlantic salmon (AS), Salmo salar, affected by amoebic gill disease (AGD). Furthermore, it explored the interaction of bithionol oral therapy with the current standard treatment
Ahmet Dobrucali et al.
World journal of gastroenterology, 10(20), 3076-3077 (2004-09-21)
Fasciola hepatica, an endemic parasite in Turkey, is still a very rare cause of cholestasis worldwide. Through ingestion of contaminated water plants like watercress, humans can become the definitive host of this parasite. Cholestatic symptoms may be sudden but in
Renee L Florent et al.
Veterinary parasitology, 144(3-4), 197-207 (2006-11-30)
This study examined the toxicity of bithionol to Atlantic salmon Salmo salar and rainbow trout Oncorhynchus mykiss in fresh- and seawater and the efficacy of bithionol as a 1h seawater bath treatment for amoebic gill disease (AGD). To examine toxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service