Skip to Content
Merck
All Photos(4)

Key Documents

C6305

Sigma-Aldrich

Clozapine

≥98% (TLC), powder, D₄-dopamine receptor antagonist

Synonym(s):

8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Clozapine,

form

powder

Quality Level

solubility

ethanol: 1 mg/mL
0.1 M HCl: 30 mg/mL
DMSO: 4.8 mg/mL

originator

Novartis

SMILES string

CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

QZUDBNBUXVUHMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Atypical antipsychotic compound. Selective antagonist for D4-dopamine receptor. Antagonist at 5-HT2A, 5-HT2C, 5-HT3, 5-HT6, and 5-HT7 serotonin receptors.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

comparable product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P B Bergqvist et al.
Psychopharmacology, 143(1), 89-96 (1999-05-05)
Low doses of the atypical antipsychotic drug risperidone are effective in patients with obsessive compulsive disorder (OCD) not responding to serotonin (5-HT) reuptake inhibitors, although higher doses have been reported to induce OCD symptoms in psychotic patients. Since such atypical
Valerio Zerbi et al.
Neuron, 103(4), 702-718 (2019-06-23)
The locus coeruleus (LC) supplies norepinephrine (NE) to the entire forebrain and regulates many fundamental brain functions. Studies in humans have suggested that strong LC activation might shift network connectivity to favor salience processing. To causally test this hypothesis, we
Owen Randlett et al.
Current biology : CB, 29(8), 1337-1345 (2019-04-09)
Habituation is a simple form of learning where animals learn to reduce their responses to repeated innocuous stimuli [1]. Habituation is thought to occur via at least two temporally and molecularly distinct mechanisms, which lead to short-term memories that last
Tarek K Rajji et al.
The American journal of psychiatry, 172(6), 579-585 (2015-04-11)
Clozapine's potent antagonism of muscarinic M1 receptors is thought to worsen working memory deficits associated with schizophrenia. In contrast, its major metabolite, N-desmethylclozapine (NDMC), is thought to enhance working memory via its M1 receptor agonist activity. The authors hypothesized that
Alexandre Picard et al.
Diabetes, 70(7), 1443-1457 (2021-04-23)
The counterregulatory response to hypoglycemia is an essential survival function. It is controlled by an integrated network of glucose-responsive neurons, which trigger endogenous glucose production to restore normoglycemia. The complexity of this glucoregulatory network is, however, only partly characterized. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service