Skip to Content
Merck
All Photos(2)

Key Documents

H0396

Sigma-Aldrich

Hydrocortisone-Water Soluble

BioReagent, suitable for cell culture

Synonym(s):

Cyclodextrin-encapsulated hydrocortisone

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
plant (wild yam)
form:
crystalline

biological source

plant (wild yam)

product line

BioReagent

form

crystalline

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 400 mg/mL

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

General description

Hydrocortisone has the potential to induce the formation of blood-brain barrier properties in serum-free culture media. It is recommended for use in epithelial and endothelial adherent cell culture applications.

Application

Hydrocortisone-Water Soluble has been used as a supplement in Dulbecco′s modified Eagle′s medium (DMEM)/high glucose medium to culture mammary alveolar (MAC-T) cells. It has also been used to study its effect on cation absorption by semicircular canal duct epithelium via epithelial sodium channel.

Packaging

Package size based on hydrocortisone

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe Perozzo Daltoe et al.
Brazilian oral research, 34, e033-e033 (2020-04-09)
The aim of our study was to isolate populations of keratinocyte stem cells based on the expression of cell surface markers and to investigate whether the culture could affect their phenotype. keratinocytes from human oral mucosa were sorted based on
Alexander Kanitz et al.
PloS one, 7(11), e49568-e49568 (2012-11-21)
Vascular endothelial growth factor A (VEGFA) plays a key role in the angiogenesis of human skin. Elevated levels of VEGFA are associated with several pathological conditions, including chronic inflammatory skin diseases and several types of skin cancer. In particular, squamous
Yohan Choi et al.
Biology of reproduction, 81(6), 1122-1130 (2009-07-31)
Calcium ions have been implicated in the establishment and maintenance of pregnancy, but the regulatory mechanisms of calcium ions in the uterine endometrium and conceptus are not well understood in pigs. Recently, we showed that TRPV6, a calcium ion channel
P H Wirtz et al.
Heart (British Cardiac Society), 90(8), 853-858 (2004-07-16)
To investigate whether stimulated monocyte cytokine release and its inhibition by glucocorticoids differs between men and women. In vitro monocyte interleukin 6 (IL-6) and tumour necrosis factor alpha (TNFalpha) release after lipopolysaccharide stimulation were assessed with and without co-incubation with
Heewon Seo et al.
Endocrinology, 149(12), 6166-6175 (2008-08-16)
Lysophosphatidic acid (LPA), a simple phospholipid-derived mediator with diverse biological actions, acts through the specific G protein-coupled receptors endothelial differentiation gene (EDG) 2, EDG4, EDG7, and GPR23. Recent studies indicate a critical role for LPA receptor signaling in embryo implantation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service