Skip to Content
Merck
All Photos(1)

Key Documents

C2755

Sigma-Aldrich

Cortisone

≥95%

Synonym(s):

17α,21-Dihydroxy-4-pregnene-3,11,20-trione, 17α-Hydroxy-11-dehydrocorticosterone, 4-Pregnene-17α,21-diol-3,11,20-trione, Kendall’s Compound E, Reichstein’s Substance Fa

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
Beilstein:
1356062
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

sterility

non-sterile

Assay

≥95%

form

powder

technique(s)

inhibition assay: suitable

mp

223-228 °C (dec.) (lit.)

solubility

chloroform: methanol (1:1): 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-15,18,22,26H,3-8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1

InChI key

MFYSYFVPBJMHGN-ZPOLXVRWSA-N

Gene Information

human ... SERPINA6(866)

Looking for similar products? Visit Product Comparison Guide

Application

Cortisone has been used:
  • to validate a 1β-HSD1 (11β-hydroxysteroid dehydrogenase type 1) inhibitor
  • to study the sensitivity of erythropoietic and granulopoietic progenitor cells to cortisone
  • in the determination of rates of cortisol oxidation by 11βHSD

Biochem/physiol Actions

Cortisone (11 dehydro 17 hydroxycortico-sterone) is an anti-inflammatory glucocorticoid that delays wound healing after surgeries. It postpones the healing process by affecting the appearance of inflammatory cells, ground substance, fibroblasts, collagen, regenerating capillaries, and epithelial migration. Glucocorticoids have anti-inflammatory, anti-allergic and immunosuppressive properties and are extensively used in treatment and therapy.
Cortisone is a glucocorticoid; a corticosterone analog that has approximately twice the anti-inflammatory potency as corticosterone but much lower Na2+ retention potency.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S McMullen et al.
Reproduction (Cambridge, England), 127(6), 717-725 (2004-06-04)
In the placenta, cortisol is inactivated by NADP(+)- and NAD(+)-dependent isoforms of 11beta-hydroxysteroid dehydrogenase (11betaHSD). Decreased placental 11betaHSD activities have been implicated in intrauterine growth restriction (IUGR) and fetal programming of adult diseases. The objective of this study was to
B Edwin et al.
Surgical endoscopy, 15(6), 589-591 (2001-10-10)
[corrected] We set out to record the operative times of an experienced laparoscopic team and assess the feasibility of outpatient laparoscopic adrenalectomy when optimal anesthesia was also offered to all patients. The study included 13 patients with aldosterone/cortisone hypersecretion and/or
Jean-Sébastien Raul et al.
Clinical biochemistry, 37(12), 1105-1111 (2004-12-14)
Since the 1960s, glucocorticoids are used by athletes to improve their performances. Their use is restricted in sports. Hair can document chronic abuse and can be therefore a complementary matrix for doping control. We have developed a new extraction, purification
F Zalman et al.
The Journal of experimental medicine, 149(1), 67-72 (1979-01-01)
The sensitivity of erythropoietic (BFU-E) and granulopoietic (CFU-C) progenitor cells to dexamethasone and cortisone was studied in cultures of mouse bone marrow. Although the log dose-response relationships had a similar form, the BFU-E were much more sensitive than the CFU-C
Hengmiao Cheng et al.
Bioorganic & medicinal chemistry letters, 20(9), 2897-2902 (2010-04-07)
The design and development of a series of highly selective pyrrolidine carboxamide 11beta-HSD1 inhibitors are described. These compounds including PF-877423 demonstrated potent in vitro activity against both human and mouse 11beta-HSD1 enzymes. In an in vivo assay, PF-877423 inhibited the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service