Skip to Content
Merck
All Photos(3)

Key Documents

C0926

Sigma-Aldrich

(2-Hydroxypropyl)-β-cyclodextrin

powder, BioReagent, suitable for cell culture

Synonym(s):

2-hydroxypropylether, Beta-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.77

biological source

corn starch

product line

BioReagent

form

powder

mol wt

1396 Da

extent of labeling

4-10 (determined by NMR)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

SMILES string

CC(O)COCC1OC2OC3C(COCC(C)O)OC(OC4C(COCC(C)O)OC(OC5C(COCC(C)O)OC(OC6C(COCC(C)O)OC(OC7C(COCC(C)O)OC(OC8C(COCC(C)O)OC(OC1C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C8OCC(C)O)C(OCC(C)O)C7OCC(C)O)C(OCC(C)O)C6OCC(C)O)C(OCC(C)O)C5OCC(C)O)C(OCC(C)O)C4OCC(C)O)C(OCC(C)O)C3OCC(C)O

InChI

1S/C63H112O42/c1-22(64)8-85-15-29-50-36(71)43(78)57(92-29)100-51-30(16-86-9-23(2)65)94-59(45(80)38(51)73)102-53-32(18-88-11-25(4)67)96-61(47(82)40(53)75)104-55-34(20-90-13-27(6)69)98-63(49(84)42(55)77)105-56-35(21-91-14-28(7)70)97-62(48(83)41(56)76)103-54-33(19-89-12-26(5)68)95-60(46(81)39(54)74)101-52-31(17-87-10-24(3)66)93-58(99-50)44(79)37(52)72/h22-84H,8-21H2,1-7H3/t22?,23?,24?,25?,26?,27?,28?,29-,30-,31?,32?,33?,34?,35?,36?,37-,38?,39-,40+,41+,42+,43?,44+,45?,46+,47+,48+,49+,50+,51+,52-,53-,54-,55-,56-,57+,58-,59+,60-,61-,62-,63-/m1/s1

InChI key

ODLHGICHYURWBS-RYJYQAAZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclodextrins are cyclic oligosaccharides consisting of 6, 7, or 8 glucopyranose units, usually referred to as α-, β-, or γ-cyclodextrins, respectively. These compounds have rigid doughnut-shaped structures making them natural complexing agents. The unique structures of these compounds owe their stability to intramolecular hydrogen bonding between the C2- and C3-hydroxyl groups of neighboring glucopyranose units. The molecule takes on the shape of a torus with the C2- and C3-hydroxyls located around the larger opening and the more reactive C6-hydroxyl aligned around the smaller opening. The arrangement of C6-hydroxyls opposite the hydrogen bonded C2- and C3-hydroxyls forces the oxygen bonds into close proximity within the cavity, leading to an electron rich, hydrophobic interior. The size of this hydrophobic cavity is a function of the number of glucopyranose units forming the cyclodextrin.

The solubility of natural cyclodextrins is very poor. In the late 1960′s, it was discovered that chemical substitutions at the 2, 3, and 6 hydroxyl sites would greatly increase solubility. Most chemically modified cyclodextrins are able to achieve a 50% (w/v) concentration in water.

Cavity size is the major determinant as to which cyclodextrin is used in complexation. The cavity diameter of β-cyclodextrins or β-glucopyranose unit compounds is well-suited for use with molecules the size of hormones, vitamins and many compounds frequently used in tissue and cell culture applications. For this reason, β-cyclodextrin is most commonly used as a complexing agent.

Application

The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.

Preparation Note

Solutions may be stored for several months at 4°C. Solid should be stored tightly sealed at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>752.0 °F

Flash Point(C)

> 400 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge L Rodriguez-Gil et al.
Human molecular genetics, 30(24), 2456-2468 (2021-07-24)
The rare, fatal neurodegenerative disorder Niemann-Pick disease type C1 (NPC1) arises from lysosomal accumulation of unesterified cholesterol and glycosphingolipids. These subcellular pathologies lead to phenotypes of hepatosplenomegaly, neurological degeneration and premature death. The timing and severity of NPC1 clinical presentation
Nicolas Schall et al.
Frontiers in pharmacology, 12, 752194-752194 (2021-11-09)
Dysregulation of CXCL12/SDF-1-CXCR4/CD184 signaling is associated with inflammatory diseases and notably with systemic lupus erythematosus. Issued from the lead molecule chalcone-4, the first neutraligand of the CXCL12 chemokine, LIT-927 was recently described as a potent analogue with improved solubility and
Melissa A Moody et al.
Molecular reproduction and development, 84(5), 408-422 (2017-04-19)
Sperm must undergo capacitation to become fertilization competent. Here we validated that monosialotetrahexosylganglioside (G
Aki Fujiwara et al.
Healthcare (Basel, Switzerland), 9(5) (2021-05-01)
The aim of this research was to investigate the antistress effect of press tack needle (PTN) acupuncture treatment using rats with social isolation stress (SIS). Rats were divided into non-stress group (Grouped+sham), stress group (SIS+sham), and PTN-treated SIS group (SIS+PTN).
Valentino J Stella et al.
Toxicologic pathology, 36(1), 30-42 (2008-03-14)
beta-cyclodextrin (beta-CD) and other cyclodextrins (CDs) have utility for solubilizing and stabilizing drugs; however, some are nephrotoxic when administered parenterally. A number of workers have attempted to identify, prepare, and evaluate various CD derivatives with superior inclusion complexation and maximal

Articles

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

Cyclodextrin solubilizes fat-soluble vitamins and hormones, enhancing their solubility in water for cell culture applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service