Skip to Content
Merck
All Photos(1)

Documents

T6397

Sigma-Aldrich

3,3′,5-Triiodo-L-thyronine sodium salt

powder, BioReagent, suitable for cell culture

Synonym(s):

Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11I3NNaO4
CAS Number:
Molecular Weight:
672.96
Beilstein:
8179867
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

product line

BioReagent

Assay

≥95%

form

powder

potency

0.02-50 ng/mL (lit.)

technique(s)

cell culture | mammalian: suitable

solubility

ethanol/1 M HCl (4:1): 10 mg/mL, clear, colorless to yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C([O-])=O

InChI

1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1

InChI key

SBXXSUDPJJJJLC-YDALLXLXSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

3,3′,5′-Triiodo-L-thyronine (T3) is an endocrine hormone produced by the thyroid gland and is regarded as an active thyroid hormone at cellular level. Thyroxine (T4) serves as a precursor for T3 and is generated in extrathyroidal tissues.

Application

3,3′,5-Triiodo-L-thyronine sodium salt has been used for inducing thyroid hormone manipulations in mice. It has also been used In cell culturing of hippocampal, retinal, proximal tubule and glomerular explant.

Biochem/physiol Actions

3,3′,5′-Triiodo-L-thyronine (T3) increases the rates of protein synthesis, stimulates the breakdown of cholesterol and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression. The interaction of T3 with nonhistone proteins in the chromatin initiates the thyroid hormone activity. Thyroid hormones regulate the differentiation and development of tissues and affect a number of metabolic processes. The level of circulating T3 hormone is reduced in hypothyroidism.

Physical form

powder-20 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw, solution stable 2-8 °C, 30 days

Reconstitution

To prepare 20 μg/ml stock solution: add 1ml 1N NaOH per mg 3,3′, 5-triiodo-L-thyronine; gently swirl to dissolve. To this, add 49 ml sterile medium per ml 1N NaOH added.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Altered focal adhesion regulation correlates with cardiomyopathy in mice expressing constitutively active rac1
Sussman MA, et al.
The Journal of Clinical Investigation, 105(7), 875-886 (2000)
Major histocompatibility complex (MHC) class I gene expression in single neurons of the central nervous system: differential regulation by interferon (IFN)-gamma and tumor necrosis factor (TNF)-alpha
Neumann H, et al.
The Journal of Experimental Medicine, 185(2), 305-316 (1997)
Hormones Growth Factors & Oncogenes, 219-219 (1987)
Handbook of Behavior, Food and Nutrition, 2109-2109 (2011)
Iván Fernández-Lamo et al.
The European journal of neuroscience, 30(4), 679-692 (2009-08-19)
Activity-dependent changes taking place at the hippocampal perforant pathway-dentate gyrus synapse during classical eyeblink conditioning were recorded in adult thyroidectomized (hypothyroid) and control (euthyroid) rats, and in animals treated with thyroid hormones 20 days after thyroidectomy (recovery rats). The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service