Skip to Content
Merck
All Photos(1)

Key Documents

P7556

Sigma-Aldrich

Progesterone-Water Soluble

powder, BioReagent, suitable for cell culture

Synonym(s):

Progesterone: HBC complex, Progesterone: methyl-β-cyclodextrin complex

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

product line

BioReagent

Assay

≥65% (HPLC)

form

powder

concentration

≥65% (HPLC)

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 25 mg/mL, clear to slightly hazy

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])C(C)=O

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Progesterone is a steroid hormone secreted by the ovaries.[1]

Application

Progesterone-Water Soluble has been used:
  • for in vitro maturation of oocyte complexes[2]
  • as a σ1R antagonist to study its effect on sigma 1 receptor[3]
  • to study its effect on the differentiation of human multipotent mesenchymal stromal cells (hMSCs) in vitro[4]

This product is not recommended for use in binding assays.

Biochem/physiol Actions

Progesterone is a hormone that maintains pregnancy. It is essential for embryo formation, development and survival in almost all mammals. This hormone exhibits its action via progesterone receptor, which is specifically expressed in the endometrial stroma throughout pregnancy.[5] It is known to prevent preterm delivery, when administered intramuscularly.[6] Progesterone also controls gonadotropin secretion, activates sexual behavior and triggers ovarian events leading to ovulation. It stimulates the growth and differentiation of the uterus and prevents uterine contractility.[7]

Packaging

Package size based on progesterone content

Other Notes

Contains approx. 70 mg progesterone per gram; balance methyl-β-cyclodextrin.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligands exert Biased Activity to regulate Sigma 1 Receptor interactions with cationic TRPA1, TRPV1 and TRPM8 channels
Cortes ME, et al.
Frontiers in Pharmacology, 10(9), 634-634 (2019)
Neurosteroid Hormones Modulate the Differentiation of Adult Human Multipotent Mesenchymal Stromal Cells
Tiftikcioglu BI, et al.
Journal of stem cell research & therapy, 4(9), 2-2 (2012)
Clinical Reproductive Medicine and Surgery, 838, 17-17 (2007)
Adding hormones sequentially could be an effective approach for IVM of dog oocytes
Evecen M, et al.
Theriogenology, 75(9), 1647-1651 (2011)
Biology of progesterone action during pregnancy recognition and maintenance of pregnancy.
Spencer T. E. and F. W. Bazer
Frontiers in Bioscience, 838, d1879-d1898 (2002)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service