Skip to Content
Merck
All Photos(1)

Documents

M-157

Supelco

Methylhexanamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17N · HCl
CAS Number:
Molecular Weight:
151.68
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCC(CC(C)N)C.Cl

InChI

1S/C7H17N.ClH/c1-4-6(2)5-7(3)8;/h6-7H,4-5,8H2,1-3H3;1H

InChI key

ZKKBPHUAHARETG-UHFFFAOYSA-N

General description

DMAA, also known as dimethylamylamine, is a stimulant sold as a dietary supplement under trade names including Geranamine and Floradrene. Since late 2009 DMAA has emerged as a club/designer drug in party pills and bath salt blends. Many professional and amateur sports bodies, such as the World Anti-Doping Agency (WADA), have banned methylhexanamine as a performance-enhancing substance. This certified solution standard is suitable for a variety of testing applications, from clinical toxicology, urine drug testing, and forensic analysis to sports testing and analysis of DMAA-based dietary supplements.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service