Skip to Content
Merck
All Photos(1)

Key Documents

R0531

Sigma-Aldrich

Ramatroban

≥98% (HPLC), solid

Synonym(s):

(+)-(3R)-3-[[(4-Fluorophenyl)sulfonyl]amino]-1,2,3,4-tetrahydro-9H-carbazole-9-propanoic acid, BAY u3405

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21FN2O4S
CAS Number:
Molecular Weight:
416.47
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ≥40 mg/mL
H2O: insoluble

originator

Bayer

storage temp.

2-8°C

SMILES string

OC(=O)CCn1c2CC[C@H](Cc2c3ccccc13)NS(=O)(=O)c4ccc(F)cc4

InChI

1S/C21H21FN2O4S/c22-14-5-8-16(9-6-14)29(27,28)23-15-7-10-20-18(13-15)17-3-1-2-4-19(17)24(20)12-11-21(25)26/h1-6,8-9,15,23H,7,10-13H2,(H,25,26)/t15-/m1/s1

InChI key

LDXDSHIEDAPSSA-OAHLLOKOSA-N

Biochem/physiol Actions

Ramatroban aids in the reduction of myocardial ischemia-reperfusion injury, lipopolysaccharide-induced shock, and vagal neuroeffector transmission in the tracheal smooth muscle of experimental models. In addition, it also suppresses allergen and IgE antibody-mediated skin and nasal reactions and eosinophilia in animal models with asthma.
Ramatroban is used for the treatment of allergic rhinitis as an antagonist of the thromboxane receptor.
Ramatroban is used for the treatment of allergic rhinitis as an antagonist of the thromboxane receptor. It is also an antagonist of the DP2 receptor with IC50 values of 100-170 nM. It is more potent at the DP2 receptor than the TP receptor by 4-5 fold. It is therefore a useful tool for the elucidation of PGD2/DP2 function in eosinophils, basophils, and other cells of the TH2 cell-type.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Royer et al.
European journal of clinical investigation, 38(9), 663-671 (2008-10-08)
Chemoattractant receptor homologous molecule of Th2 cells (CRTH2) has been shown to mediate the chemotaxis of eosinophils, basophils and Th2-type T lymphocytes. The major mast cell product prostaglandin (PG) D(2) is considered to be the principal ligand of CRTH2. We
Eduardo Busto et al.
The Journal of organic chemistry, 77(10), 4842-4848 (2012-04-21)
A chemoenzymatic asymmetric route for the preparation of enantiopure (R)-ramatroban has been developed for the first time. The action of lipases and oxidoreductases has been independently studied, and both were found as excellent biocatalysts for the production of adequate chiral
Tetsuro Wada et al.
Auris, nasus, larynx, 33(4), 455-460 (2006-08-11)
The pathogenesis of eosinophilic otitis media is not yet fully understood. The purpose of this paper is to describe the clinical course of our two patients with eosinophilic otitis media and to discuss the pathogenesis and treatment of this intractable
Shân L Gyles et al.
Immunology, 119(3), 362-368 (2006-10-28)
Human cultured mast cells, immunologically activated with immunoglobuin E (IgE)/anti-IgE, released a factor(s) that promoted chemotaxis of human CRTH2+ CD4+ T helper type 2 (Th2) lymphocytes. Mast cell supernatants collected at 20 min, 1 hr, 2 hr and 4 hr
Update on the development of antagonists of chemoattractant receptor-homologous molecule expressed on Th2 cells (CRTH2). From lead optimization to clinical proof-of-concept in asthma and allergic rhinitis.
Roy Pettipher et al.
Journal of medicinal chemistry, 55(7), 2915-2931 (2012-01-10)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service