Skip to Content
Merck
All Photos(1)

Key Documents

A-016

Supelco

(±)-Amphetamine-d11 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-Amphetamine-d11 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D11H2N
CAS Number:
Molecular Weight:
146.27
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C9H13N/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/i1D3,2D,3D,4D,5D,6D,7D2,8D

InChI key

KWTSXDURSIMDCE-AAFMOIDSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall®, Vyvanse®, and ProCentra®. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This stable-labeled internal standard is suitable for quantitation of amphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or sports testing.

Application


  • Development of a certified reference material: An SI-traceable N-terminal pro-B-type natriuretic peptide certified reference material was developed using structure-based impurity-corrected isotope dilution mass spectrometry approaches, which is applicable to improving the accuracy of biomarker measurements in clinical diagnostics (Xiao et al., 2024).

  • Advanced materials for bone regeneration: Research on large-scale self-mineralizing bone organoids used bone matrix-inspired hydroxyapatite hybrid bioinks, enhancing the potential of 3D bioprinting technologies for regenerative medicine and providing a foundation for further studies on biomaterials and tissue engineering (Wang et al., 2024).

  • Predictive models in prenatal care: A deep learning model was developed to predict renal anomalies in prenatal ultrasound diagnostics, demonstrating the utility of AI in enhancing diagnostic accuracy and early intervention strategies in maternal-fetal medicine (Miguel et al., 2024).

  • Environmental technology for pollution control: Lignin nanoparticles were explored as an efficient adsorbent for the removal of methylene blue from aqueous media, showing promise for applications in water treatment and environmental cleanup efforts (Pourbaba et al., 2024).

Legal Information

Adderall is a registered trademark of Richwood Pharmaceutical Company, Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
ProCentra is a registered trademark of Outlook Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyvanse is a registered trademark of Shire, LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service