Skip to Content
Merck
All Photos(1)

Key Documents

A-008

Supelco

S(+)-Amphetamine (dextro-Amphetamine) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

dextro-Amphetamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H](N)Cc1ccccc1

InChI

1S/C9H13N/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/t8-/m0/s1

InChI key

KWTSXDURSIMDCE-QMMMGPOBSA-N

Gene Information

human ... SLC18A2(6571)

General description

Amphetamine is a stimulant of the phenethylamine class found in ADHD-treatment drugs such as Adderall®, Vyvanse®, and ProCentra®. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This Certified Snap-N-Spike® Solution is suitable for use in urine drug testing, clinical toxicology, forensic analysis, and isomer identification by LC-MS/MS or GC/MS.

Legal Information

Adderall is a registered trademark of Richwood Pharmaceutical Company, Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
ProCentra is a registered trademark of Outlook Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyvanse is a registered trademark of Shire, LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L J Schouw et al.
NeuroImage, 72, 1-9 (2013-01-09)
Pharmacological magnetic resonance imaging (phMRI) maps the neurovascular response to a pharmacological challenge and is increasingly used to assess neurotransmitter systems. Here we investigated the hemodynamic response to a dopaminergic (DAergic) challenge with dextroamphetamine (dAMPH) in humans using arterial spin
Amanda V Steckert et al.
Neurochemistry international, 62(4), 425-432 (2013-02-16)
Several evidences have demonstrated that oxidative stress has a central role in bipolar disorder (BD). Recently, studies have been suggested histone deacetylases (HDAC) as a possible target for new medications in treatment of mood disorders. In this study, we investigated
Greg W Mattingly et al.
BMC psychiatry, 13, 39-39 (2013-01-30)
Despite the overall high degree of response to pharmacotherapy, consensus is lacking on how to judge clinical response or define optimal treatment/remission when treating adults with attention-deficit/hyperactivity disorder (ADHD). This study examined clinical response and symptomatic remission in analyses of
Joel L Young
Postgraduate medicine, 125(1), 162-168 (2013-02-09)
Fatigue is commonly reported in the primary care setting; however, its cause is often unclear. This article presents 3 cases involving patients with chronic fatigue syndrome who responded poorly to treatment. After clinical evaluation, all patients were found to meet
J H Check et al.
Clinical and experimental obstetrics & gynecology, 40(2), 227-228 (2013-08-27)
To further investigate the efficacy of treatment of interstitial cystitis that had been refractory to standard treatment with sympathomimetic amines. Dextroamphetamine sulfate sustained release capsules up to 30 mg per day were prescribed in women with refractory painful bladder syndrome/interstitial

Protocols

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service