Skip to Content
Merck
All Photos(1)

Key Documents

160962

Sigma-Aldrich

Succinonitrile

99%

Synonym(s):

1,2-Dicyanoethane, Butanedinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CH2CN
CAS Number:
Molecular Weight:
80.09
Beilstein:
1098380
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

265-267 °C (lit.)

mp

50-54 °C (lit.)

solubility

methanol: soluble 1 g/10 mL, clear, colorless
water: soluble 130 g/L

density

0.985 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

N#CCCC#N

InChI

1S/C4H4N2/c5-3-1-2-4-6/h1-2H2

InChI key

IAHFWCOBPZCAEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Succinonitrile has plastic crystal nature. It is the precursor of 1,4-diaminobutane.

Application

Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jahaziel Villarreal et al.
Membranes, 10(3) (2020-03-21)
In the present work, the effect of temperature and additives on the ionic conductivity of mixed organic/ionic liquid electrolytes (MOILEs) was investigated by conducting galvanostatic charge/discharge and ionic conductivity experiments. The mixed electrolyte is based on the ionic liquid (IL)
A Combined Conductivity and XAS Study of Plastically Crystalline Electrolytes.
Chadwick AV, et al.
Journal of Physics. Conference Series, 249(1) (2010)
Song Wang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 81(11), 2311-2321 (2020-08-14)
As porous crystal materials, metal-organic frameworks (MOFs) have attracted wide attention in the field of environmental remediation. In this study, a trivalent iron-tartaric acid metal-organic framework (T2-MOF) was successfully synthesized using the inexpensive raw materials ferric chloride (FeCl3.6H2O) and tartaric
Studies on the systems involved in succinonitrile metabolism in liver.
M Floreani et al.
Pharmacological research communications, 12(5), 433-439 (1980-05-01)
Mohd Suleman et al.
The journal of physical chemistry. B, 117(24), 7436-7443 (2013-06-14)
Experimental studies on the novel compositions of gel polymer electrolytes, comprised of plastic crystal succinonitrile (SN) dispersed with pyrrolidinium and imidazolium-based ionic liquids (ILs) entrapped in a host polymer poly(vinylidine fluoride-co-hexafluoropropylene) (PVdF-HFP), are reported. The gel electrolytes are in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service