Skip to Content
Merck
All Photos(1)

Key Documents

114588

Sigma-Aldrich

5-(4-Dimethylaminobenzylidene)rhodanine

97%

Synonym(s):

5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one, p-(Dimethylamino)benzal-5-rhodanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2OS2
CAS Number:
Molecular Weight:
264.37
Beilstein:
189065
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

technique(s)

titration: suitable

mp

285-288 °C (lit.)

solubility

H2O: insoluble
acetone: moderately soluble
benzene: very slightly soluble
chloroform: very slightly soluble
diethyl ether: very slightly soluble

λmax

451 nm

SMILES string

CN(C)c1ccc(cc1)\C=C2\SC(=S)NC2=O

InChI

1S/C12H12N2OS2/c1-14(2)9-5-3-8(4-6-9)7-10-11(15)13-12(16)17-10/h3-7H,1-2H3,(H,13,15,16)/b10-7+

InChI key

JJRVRELEASDUMY-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-(4-Dimethylaminobenzylidene)rhodanine is a silver-specific dye that is used to quantitate the autoradiographic deposition of silver onto X-ray film in a colorimetric assay. 5-(4-Dimethylaminobenzylidene)rhodanine is used as an indicator in the titration of cyanide solution with silver nitrate solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casassas E, et al.
Analyst, 109(9), 1159-1163 (1984)
P Emanuele et al.
Annals of diagnostic pathology, 2(2), 125-126 (1998-12-10)
We report a modified copper stain that can be completed in 2 hours, rather than overnight, which is required for other stains. This is useful in the diagnosis of chronic cholestatic syndromes, Wilson's disease, Indian childhood cirrhosis, and other conditions
J W Ludlow et al.
Analytical biochemistry, 154(1), 104-109 (1986-04-01)
5-(4-Dimethylaminobenzylidene)rhodamine, a silver-specific dye, was used in a colorimetric assay to quantitate the autoradiographic deposition of silver onto X-ray film after exposure to sodium dodecyl sulfate-polyacrylamide gels of radiolabeled biological material. Silver grains were eluted from autoradiograms with 5 N
J S Kumaratilake et al.
Research in veterinary science, 42(1), 73-81 (1987-01-01)
Six Merino sheep were dosed orally with a 0.2 per cent solution of copper sulphate, six others were undosed controls. Liver biopsies were obtained and stained for copper by the p-dimethylaminobenzylidene rhodanine (DMABR), rubeanic acid (RA) and ferricyanide (FCN) methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service