Skip to Content
Merck
All Photos(1)

Key Documents

14190

Sigma-Aldrich

Succinonitrile

purum, ≥97.0% (GC)

Synonym(s):

1,2-Dicyanoethane, Butanedinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CH2CN
CAS Number:
Molecular Weight:
80.09
Beilstein:
1098380
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (GC)

form

solid

bp

265-267 °C (lit.)

mp

50-54 °C (lit.)

solubility

water: soluble 130 g/L

density

0.985 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

N#CCCC#N

InChI

1S/C4H4N2/c5-3-1-2-4-6/h1-2H2

InChI key

IAHFWCOBPZCAEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Succinonitrile has plastic crystalline nature. It is the precursor of 1,4-diaminobutane.

Application

Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd C Peterson et al.
Journal of neurotrauma, 29(18), 2823-2830 (2012-09-29)
The primary goal of this study was to compare clinically relevant doses of progesterone and nicotinamide within the same injury model. Progesterone has been shown to reduce edema and inflammation and improve functional outcomes following brain injury. Nicotinamide has also
A Combined Conductivity and XAS Study of Plastically Crystalline Electrolytes.
Chadwick AV, et al.
Journal of Physics. Conference Series, 249(1) (2010)
Possible role of hydroxyl radicals in the metabolism of succinonitrile.
E P Hayes et al.
Biochemical pharmacology, 34(22), 4081-4084 (1985-11-15)
O I Fengler et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(1), 105-117 (2001-02-24)
[1,4-13C2]-succinonitrile, [2,2,3,3-2H4]-succinonitrile, [1,4-13C2-2,2,3,3-2H4]-succinonitrile have been synthesized and, for the first time, the infrared and Raman spectra of these succinonitrile isotopomers have been discussed in detail. The spectra were recorded at ambient temperature and at the temperature of liquid nitrogen and
Mohd Suleman et al.
The journal of physical chemistry. B, 117(24), 7436-7443 (2013-06-14)
Experimental studies on the novel compositions of gel polymer electrolytes, comprised of plastic crystal succinonitrile (SN) dispersed with pyrrolidinium and imidazolium-based ionic liquids (ILs) entrapped in a host polymer poly(vinylidine fluoride-co-hexafluoropropylene) (PVdF-HFP), are reported. The gel electrolytes are in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service