Skip to Content
Merck
All Photos(3)

Key Documents

D77001

Sigma-Aldrich

Adiponitrile

99%

Synonym(s):

1,6-hexanedinitrile, ADN, 1,4-Dicyanobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NC(CH2)4CN
CAS Number:
Molecular Weight:
108.14
Beilstein:
1740005
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 40 °C)

Assay

99%

form

liquid

autoignition temp.

1022 °F

expl. lim.

4.99 %

refractive index

n20/D 1.438 (lit.)

bp

295 °C (lit.)

mp

1-3 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

N#CCCCCC#N

InChI

1S/C6H8N2/c7-5-3-1-2-4-6-8/h1-4H2

InChI key

BTGRAWJCKBQKAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Adiponitrile is a key precursor for manufacturing nylon-6,6. It can undergo catalytic hydrogenation to form hexamethylenediamine (HMD). ADN shows promising potential as a solvent for electrolyte solutions used in high-voltage lithium batteries due to its high anodic and thermal stability.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

325.4 °F - closed cup

Flash Point(C)

163 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raghuraj Rao et al.
Analytica chimica acta, 599(1), 24-35 (2007-09-04)
Multivariate calibration problems often involve the identification of a meaningful subset of variables, from a vast number of variables for better prediction of output variables. A new graph theoretic method based on partial correlations (variable interaction network-VIN) is proposed. Many
Jian-Jun Dai et al.
ChemSusChem, 5(4), 617-620 (2012-03-24)
Current affairs: Adiponitrile, used to produce nylon 6.6, is prepared from the renewable compound glutamic acid by an electrochemical route, involving electro-oxidative decarboxylation and Kolbe coupling reactions. The new route is an example of the use of glutamic acid as
O L Dashiell et al.
Journal of applied toxicology : JAT, 4(6), 320-325 (1984-12-01)
Nine chemicals, with a range from extremely to slightly toxic, were used to measure the oral LD50 in both fasted (24-h) and non-fasted rats. Each chemical was tested as a solution or suspension in corn oil, responses within 14 days
Inhalation toxicity of adiponitrile in rats.
L W Smith et al.
Toxicology and applied pharmacology, 65(2), 257-263 (1982-09-15)
J L Moreau et al.
Journal of chromatography. B, Biomedical applications, 656(1), 197-202 (1994-06-03)
A procedure for the assay of nitrile hydratase and amidase activity by high-performance liquid chromatography is described. The method can be used to assay the intermediate compounds resulting from the hydrolysis of adiponitrile into adipic acid, and to determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service