Skip to Content
Merck
All Photos(3)

Key Documents

757136

Sigma-Aldrich

Lithium bis(oxalato)borate

greener alternative

Synonym(s):

LiBOB, Lithium bis(ethanedioato)borate, Lithium bis(oxalate)borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiB(C2O4)2
CAS Number:
Molecular Weight:
193.79
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

powder or crystals

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].O=C1O[B-]2(OC1=O)OC(=O)C(=O)O2

InChI

1S/C4BO8.Li/c6-1-2(7)11-5(10-1)12-3(8)4(9)13-5;/q-1;+1

InChI key

NVQAYVUCVASGDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium bis(oxalato)borate (LiBOB) is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LiBOB is a novel boron based Li salt electrolyte material for Li ion batteries. It is environmentally friendly with good film forming property and high thermal stability and is compatible with a variety of anodes and metal oxide cathode.
LiBOB is a thermally stable electrolyte that can be used to protect graphite-based anode materials in lithium-ion batteries. It shows good electrochemical performance with a discharge capacity retention of ~ 83%.

Legal Information

Product of Albemarle US Inc

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium bis (oxalato) borate stabilizes graphite anode in propylene carbonate
Xu K, et al.
Electrochemical and Solid-State Letters, 5(11), A259-A259 (2002)
Shoichi Matsuda et al.
Scientific reports, 9(1), 6211-6211 (2019-04-19)
Data-driven material discovery has recently become popular in the field of next-generation secondary batteries. However, it is important to obtain large, high quality data sets to apply data-driven methods such as evolutionary algorithms or Bayesian optimization. Combinatorial high-throughput techniques are
Wengao Zhao et al.
ChemSusChem, 11(13), 2211-2220 (2018-05-03)
The long-term cycling performance, rate capability, and voltage stability of lithium (Li) metal batteries with LiNi0.76 Mn0.14 Co0.10 O2 (NMC76) cathodes is greatly enhanced by lithium bis(oxalato)borate (LiBOB) additive in the LiPF6 -based electrolyte. With 2 % LiBOB in the electrolyte
R Verrelli et al.
Physical chemistry chemical physics : PCCP, 19(38), 26435-26441 (2017-09-26)
Layered MgMoN
Jenny Strehlau et al.
Analytical and bioanalytical chemistry, 409(26), 6123-6131 (2017-08-05)
A novel method based on liquid-liquid extraction with subsequent gas chromatography separation and mass spectrometric detection (GC-MS) for the quantification of organic carbonates in cell culture materials is presented. Method parameters including the choice of extraction solvent, of extraction method

Articles

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service