Merck
Plasma and cerebrospinal fluid amino acids in epileptic patients.
Neurochemical research (2004-03-03)
Sirpa Rainesalo, Tapani Keränen, Johanna Palmio, Jukka Peltola, Simo S Oja, Pirjo Saransaari
ABSTRACT
Altered plasma and cerebrospinal fluid amino acid levels may be associated with human epilepsy. We studied three groups of patients, those with a generalized epileptic syndrome, juvenile myoclonic epilepsy, patients with refractory localization-related epilepsies, and patients with acute seizures (within 24 h). Plasma levels of amino acids were studied in all patient groups, as were those in the cerebrospinal fluid (CSF) of patients with acute seizures. After acute seizures, the amino acid changes in the CSF were limited to a reduction in the level of taurine, whereas the levels of most amino acids in plasma were decreased. On the other hand, levels of the excitatory amino acids glutamate and aspartate were increased. The most notable finding in the juvenile myoclonic epilepsy patients was an increase in glutamate level in the plasma. Our study supports the conception of an altered metabolism of glutamate in generalized epilepsies.
Product Description
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Cystine
SAFC
L-Aspartic acid
SAFC
L-Valine
SAFC
L-Threonine
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
SAFC
L-Tyrosine
Alanine, European Pharmacopoeia (EP) Reference Standard
Cystine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
SAFC
Glycine
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)