Skip to Content
Merck
All Photos(1)

Key Documents

76526

Supelco

L-Leucine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
E Number:
E641
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

20-25°C

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Leucine is an important and essential branched chain amino acid. It shows activity with the mechanistic target of rapamycin(mTOR) pathway, involved in the synthesis of proteins and satiety.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mullin.EG, et al. et al.
Integrative Weight Management: A Guide for Clinicians (2014)
Fleck M and Petrosyan.MA et al.
Salts of Amino Acids: Crystallization, Structure and Properties (2014)
Michael J Ormsby et al.
Cell reports, 30(7), 2297-2305 (2020-02-23)
Propionic acid (PA) is a bacterium-derived intestinal antimicrobial and immune modulator used widely in food production and agriculture. Passage of Crohn's disease-associated adherent-invasive Escherichia coli (AIEC) through a murine model, in which intestinal PA levels are increased to mimic the
T Yoshimasa et al.
The Journal of clinical investigation, 69(3), 643-650 (1982-03-01)
To elucidate the physiological and pathophysiological significance of methionine- and leucine-enkephalin (Met-and Leu-enkephalin, respectively) in human sympathoadrenal system, the contents of these peptides in normal human sympathetic nervous system, adrenal medulla, and pheochromocytomas were determined by specific radioimmunoassays combined with
William I A Haynes et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(11), 4804-4814 (2013-03-15)
The identification of a hyperdirect cortico-subthalamic nucleus connection highlighted the important role of the subthalamic nucleus (STN) in regulating behavior. However, this pathway was shown primarily from motor areas. Hyperdirect pathways associated with cognitive and motivational cortical regions are particularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service