12-1210
Glycine
SAJ special grade, ≥99.0%
Synonym(s):
Aminoacetic acid, Aminoethanoic acid, Glycocoll
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
grade:
SAJ special grade
Assay:
≥99.0%
application(s):
peptide synthesis
Recommended Products
grade
SAJ special grade
Assay
≥99.0%
form
crystalline powder
availability
available only in Japan
pH
5.9-6.4 (50 g/L)
pKa (25 °C)
(1) 2.35, (2) 9.60
2.35
mp
240 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
NCC(O)=O
InChI
1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)
InChI key
DHMQDGOQFOQNFH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Richard A Felix et al.
Brain structure & function, 220(5), 2639-2652 (2014-06-30)
The mammalian superior paraolivary nucleus (SPON) is a major source of GABAergic inhibition to neurons in the inferior colliculus (IC), a well-studied midbrain nucleus that is the site of convergence and integration for the majority ascending auditory pathways en route
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Li-Chun Tu et al.
The EMBO journal, 32(24), 3220-3230 (2013-11-12)
Nuclear pore complexes (NPCs) mediate cargo traffic between the nucleus and the cytoplasm of eukaryotic cells. Nuclear transport receptors (NTRs) carry cargos through NPCs by transiently binding to phenylalanine-glycine (FG) repeats on intrinsically disordered polypeptides decorating the NPCs. Major impediments
Suzanne L Grant et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 844(2), 278-282 (2006-08-08)
A simple and versatile methodology using high-performance liquid chromatography (HPLC) with fluorimetric detection was developed to simultaneously determine d-serine along with other metabolically related neuroactive amino acids in the glutamatergic system: L-serine, L-glutamate, L-glutamine, and glycine. On-column sensitivity was in
Zoé Husson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(28), 9418-9431 (2014-07-11)
The principal neurons of the cerebellar nuclei (CN), the sole output of the olivo-cerebellar system, receive a massive inhibitory input from Purkinje cells (PCs) of the cerebellar cortex. Morphological evidence suggests that CN principal cells are also contacted by inhibitory
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service