Skip to Content
Merck
All Photos(1)

Key Documents

C5735

SAFC

L-Cystine

Synonym(s):

(R,R)-3,3′-Dithiobis(2-aminopropionicacid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein:
1728094
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

biological source

non-animal source

Assay

≥98.5%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

mp

>240 °C (dec.) (lit.)

solubility

1 M HCl: 100 mg/mL

suitability

suitable for manufacturing use

application(s)

pharmaceutical (small molecule)

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip J Hogg
Nature reviews. Cancer, 13(6), 425-431 (2013-05-11)
Protein action in nature is generally controlled by the amount of protein produced and by chemical modification of the protein, and both are often perturbed in cancer. The amino acid side chains and the peptide and disulphide bonds that bind
Serge-George Rosolen et al.
Advances in experimental medicine and biology, 775, 145-154 (2013-02-09)
In the 70s, the amino acid taurine was found essential for photoreceptor survival. Recently, we found that taurine depletion can also trigger retinal ganglion cell degeneration both in vitro and in vivo. Therefore, evaluation of taurine levels could be a
Martijn J Wilmer et al.
American journal of physiology. Renal physiology, 299(5), F905-F916 (2010-09-10)
Renal proximal tubules are highly sensitive to ischemic and toxic insults and are affected in diverse genetic disorders, of which nephropathic cystinosis is the most common. The disease is caused by mutations in the CTNS gene, encoding the lysosomal cystine
Anne C Conibear et al.
The Journal of biological chemistry, 288(15), 10830-10840 (2013-02-23)
θ-Defensins are ribosomally synthesized cyclic peptides found in the leukocytes of some primate species and have promising applications as antimicrobial agents and scaffolds for peptide drugs. The cyclic cystine ladder motif, comprising a cyclic peptide backbone and three parallel disulfide
Jan Lewerenz et al.
Antioxidants & redox signaling, 18(5), 522-555 (2012-06-07)
The antiporter system x(c)(-) imports the amino acid cystine, the oxidized form of cysteine, into cells with a 1:1 counter-transport of glutamate. It is composed of a light chain, xCT, and a heavy chain, 4F2 heavy chain (4F2hc), and, thus

Articles

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Importance and uses of cysteine in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service