Skip to Content
Merck
All Photos(1)

Key Documents

C3300000

Cystine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Cystine, (R,R)-3,3′-Dithiobis(2-aminopropionicacid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein:
1728094
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cystine

manufacturer/tradename

EDQM

mp

>240 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cystine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne C Conibear et al.
The Journal of biological chemistry, 288(15), 10830-10840 (2013-02-23)
θ-Defensins are ribosomally synthesized cyclic peptides found in the leukocytes of some primate species and have promising applications as antimicrobial agents and scaffolds for peptide drugs. The cyclic cystine ladder motif, comprising a cyclic peptide backbone and three parallel disulfide
Jan Lewerenz et al.
Antioxidants & redox signaling, 18(5), 522-555 (2012-06-07)
The antiporter system x(c)(-) imports the amino acid cystine, the oxidized form of cysteine, into cells with a 1:1 counter-transport of glutamate. It is composed of a light chain, xCT, and a heavy chain, 4F2 heavy chain (4F2hc), and, thus
Philip J Hogg
Nature reviews. Cancer, 13(6), 425-431 (2013-05-11)
Protein action in nature is generally controlled by the amount of protein produced and by chemical modification of the protein, and both are often perturbed in cancer. The amino acid side chains and the peptide and disulphide bonds that bind
Serge-George Rosolen et al.
Advances in experimental medicine and biology, 775, 145-154 (2013-02-09)
In the 70s, the amino acid taurine was found essential for photoreceptor survival. Recently, we found that taurine depletion can also trigger retinal ganglion cell degeneration both in vitro and in vivo. Therefore, evaluation of taurine levels could be a
Ojore Benedict Valentine Oka et al.
Molecular cell, 50(6), 793-804 (2013-06-19)
ERdj5 is a member of the protein disulfide isomerase family of proteins localized to the endoplasmic reticulum (ER) of mammalian cells. To date, only a limited number of substrates for ERdj5 are known. Here we identify a number of endogenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service