Skip to Content
Merck
All Photos(5)

Documents

G7126

Sigma-Aldrich

Glycine

≥99% (HPLC), suitable for western blotting, ReagentPlus®

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

Glycine, ReagentPlus®, ≥99% (HPLC)

product line

ReagentPlus®

Assay

≥99% (HPLC)

form

powder

technique(s)

western blot: suitable

impurities

≤0.01% Ammonia

loss

≤0.2% loss on drying

color

white to off-white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

H2O: 200 mg/mL, clear, colorless to faintly yellow

cation traces

≤0.01% (Ammonia (NH3))

functional group

amine
carboxylic acid

storage temp.

room temp

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Glycine is a non-chiral amino acid. Glycine is a molecule with pI of 6.7, which is similar to the pH of stacking region in polyacrylamide gels. It has advantage of low mobility, hydrophobicity and it does not associate with proteins.
Glycine is a proteinogenic amino acid that plays a diverse array of roles in biochemistry and physiology. This non-essential, non-polar glucogenic amino acid acts as an inhibitory neurotransmitter in the central nervous system, but also works alongside glutamate as a co-agonist at NMDA receptors. As a precursor, glycine serves as a building block for various macromolecules, playing a crucial role in cellular processes.

Beyond its vital biological functions, glycine demonstrates remarkable versatility in various research applications. Its application as a biochemical reagent makes it invaluable in numerous assays and procedures. Its buffering capacity allows it to play a critical role in protein analysis techniques like SDS-PAGE and Western Blotting, chromatography, and cell culture. Glycine′s zwitterionic nature makes it an effective buffer across a range of pH values. In immunology research, it is widely used in the preparation of buffers for Western Blotting and other techniques. Additionally, its compatibility with various enzymes makes it useful in enzymatic assays like lactate determination. Furthermore, glycine contributes to the formulation of buffers for protein stabilization, pH control, and enzymatic reactions.

Application

  • Glycine has been added in the transfer buffer during western blotting procedure.
  • It has been used in the solution prepared for dissolving formazan crystals in the MTT (3-[4,5-dimethylthiazol-2-yl]-2,5 diphenyltetrazolium bromide) cytotoxicity assay.
  • It has been used for terminating the rat basophilic leukemia (RBL) assay.

Biochem/physiol Actions

Glycine is an inhibitory neurotransmitter in the central nervous system and works as an allosteric regulator of NMDA (N-methyl-D-aspartate) receptors. It is involved in processing of motor and sensory data, thereby regulating movement, vision and audition.
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Features and Benefits

  • Suitable for Cell Biology and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of the sensitizing potential of processed peanut proteins in Brown Norway rats: roasting does not enhance allergenicity.
Kroghsbo S et al.
PLoS ONE, 9, e96475-e96475 (2014)
A functional study on the migration of human monocytes to human leukemic cell lines and the role of monocyte chemoattractant protein-1.
Legdeur MC et al.
Leukemia, 11, 1904-1904 (1997)
Amino Acids, Peptides and Proteins in Organic Chemistry (2013)
Formalin-inactivated bovine RSV vaccine enhances a Th2 mediated immune response in infected cattle.
Kalina WV et al.
Vaccine, 22, 1465-1465 (2004)
Westermeier R
Electrophoresis in Practice: A Guide to Methods and Applications of DNA and Protein Separations (2016)

Protocols

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service