Merck
  • Genetic mapping of targets mediating differential chemical phenotypes in Plasmodium falciparum.
Genetic mapping of targets mediating differential chemical phenotypes in Plasmodium falciparum.
Nature chemical biology (2009-09-08)
Jing Yuan, Ronald L Johnson, Ruili Huang, Jennifer Wichterman, Hongying Jiang, Karen Hayton, David A Fidock, Thomas E Wellems, James Inglese, Christopher P Austin, Xin-zhuan Su
ABSTRACT
Studies of gene function and molecular mechanisms in Plasmodium falciparum are hampered by difficulties in characterizing and measuring phenotypic differences between individual parasites. We screened seven parasite lines for differences in responses to 1,279 bioactive chemicals. Hundreds of compounds were active in inhibiting parasite growth; 607 differential chemical phenotypes, defined as pairwise IC(50) differences of fivefold or more between parasite lines, were cataloged. We mapped major determinants for three differential chemical phenotypes between the parents of a genetic cross, and we identified target genes by fine mapping and testing the responses of parasites in which candidate genes were genetically replaced with mutant alleles. Differential sensitivity to dihydroergotamine methanesulfonate (1), a serotonin receptor antagonist, was mapped to a gene encoding the homolog of human P-glycoprotein (PfPgh-1). This study identifies new leads for antimalarial drugs and demonstrates the utility of a high-throughput chemical genomic strategy for studying malaria traits.
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
(−)-p-Bromolevamisole oxalate, 99%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
S-(+)-Fluoxetine hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
Harmane, 98%
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Riluzole, solid
Sigma-Aldrich
Imipramine hydrochloride, ≥99% (TLC)
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Decamethonium bromide, crystalline
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Tetramisole hydrochloride, phosphatase inhibitor
Sigma-Aldrich
Nortriptyline hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%
Sigma-Aldrich
Naltrexone hydrochloride
Sigma-Aldrich
Pancuronium bromide
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder