Skip to Content
Merck
All Photos(4)

Documents

R116

Sigma-Aldrich

Riluzole

≥98% (HPLC), solid, glutamate release inhibitor

Synonym(s):

2-Amino-6-(trifluoromethoxy)benzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5F3N2OS
CAS Number:
Molecular Weight:
234.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Riluzole, solid

form

solid

originator

Sanofi Aventis

SMILES string

Nc1nc2ccc(OC(F)(F)F)cc2s1

InChI

1S/C8H5F3N2OS/c9-8(10,11)14-4-1-2-5-6(3-4)15-7(12)13-5/h1-3H,(H2,12,13)

InChI key

FTALBRSUTCGOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glutamate release inhibitor; anticonvulsant

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan S Phillips et al.
eLife, 11 (2022-07-08)
Previously our computational modeling studies (Phillips et al., 2019) proposed that neuronal persistent sodium current (INaP) and calcium-activated non-selective cation current (ICAN) are key biophysical factors that, respectively, generate inspiratory rhythm and burst pattern in the mammalian preBötzinger complex (preBötC)
B C Cheah et al.
Current medicinal chemistry, 17(18), 1942-1199 (2010-04-10)
Amyotrophic lateral sclerosis (ALS) is a universally fatal neurodegenerative disease of the human motor system. Aetiological mechanisms implicated in the development of ALS have been linked to the glutamatergic neurotransmitter system, with destruction of motor neurons triggered through excessive activation
Steve Vucic et al.
Brain : a journal of neurology, 136(Pt 5), 1361-1370 (2013-04-26)
Riluzole, a benzothiazole derivative, has been shown to be effective in prolonging survival in amyotrophic lateral sclerosis. The mechanisms by which riluzole exerts neuroprotective effects in amyotrophic lateral sclerosis remains to be fully elucidated, although inhibition of glutamatergic transmission and
Belén Mollá et al.
Molecular neurobiology, 59(10), 6018-6032 (2022-07-15)
Lafora disease (LD) is a fatal rare neurodegenerative disorder that affects young adolescents and has no treatment yet. The hallmark of LD is the presence of polyglucosan inclusions (PGs), called Lafora bodies (LBs), in the brain and peripheral tissues. LD
Arun Pal et al.
Life science alliance, 7(8) (2024-05-18)
Amyotrophic lateral sclerosis (ALS) leads to death within 2-5 yr. Currently, available drugs only slightly prolong survival. We present novel insights into the pathophysiology of Superoxide Dismutase 1 (SOD1)- and in particular Fused In Sarcoma (FUS)-ALS by revealing a supposedly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service