Skip to Content
Merck
All Photos(1)

Documents

Q3251

Sigma-Aldrich

Quinacrine dihydrochloride

≥90% (TLC), powder, MAO-A/B inhibitor

Synonym(s):

6-Chloro-9-(4-diethylamino-1-methylbutylamino)-2-methoxyacridine dihydrochloride, Atebrin dihydrochloride, Mepacrine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30ClN3O · 2HCl
CAS Number:
Molecular Weight:
472.88
Beilstein:
4834013
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.77

product name

Quinacrine dihydrochloride, ≥90%

biological source

synthetic

Assay

≥90%

form

powder

mp

257 °C

solubility

H2O: soluble, clear to hazy

originator

Bayer

SMILES string

Cl[H].Cl[H].CCN(CC)CCCC(C)Nc1c2ccc(Cl)cc2nc3ccc(OC)cc13

InChI

1S/C23H30ClN3O.2ClH/c1-5-27(6-2)13-7-8-16(3)25-23-19-11-9-17(24)14-22(19)26-21-12-10-18(28-4)15-20(21)23;;/h9-12,14-16H,5-8,13H2,1-4H3,(H,25,26);2*1H

InChI key

UDKVBVICMUEIKS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Quinacrine is an acridine derivative and a potent antimalarial agent.

Application

Quinacrine dihydrochloride has been used:
  • in its uptake and accumulation studies in mouse lung slices using fluorescence microscope
  • in the staining of ATP vesicles in mesenchymal stem cells (MSCs)
  • in uptake-release assay for characterization of dense granule functionality of platelets

Biochem/physiol Actions

Quinacrine has anthelmintic functionality and used in the female sterilization. It is used to treat systemic lupus erythematosus and rheumatic diseases. Apart from malaria, Quinacrine is effective for treating giardia and tapeworm infections. It inhibits nicotinamide adenine dinucleotide phosphate (NADPH) oxidase activity in addition to monoamine oxidase inhibition.
Non-selective MAO-A/B inhibitor.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abnormal differentiation of B cells and megakaryocytes in patients with Roifman syndrome
Heremans J, et al.
The Journal of Allergy and Clinical Immunology, 142(2), 630-646 (2018)
Denise De Zanet et al.
PloS one, 12(9), e0184941-e0184941 (2017-09-19)
A new biosensor for the real-time analysis of thrombus formation is reported. The fast and accurate monitoring of the individual thrombotic risk represents a challenge in cardiovascular diagnostics and in treatment of hemostatic diseases. Thrombus volume, as representative index of
Mayur Nimbadas Devare et al.
Aging cell, 19(6), e13151-e13151 (2020-05-26)
Glucose controls the phosphorylation of silent information regulator 2 (Sir2), a NAD+ -dependent protein deacetylase, which regulates the expression of the ATP-dependent proton pump Pma1 and replicative lifespan (RLS) in yeast. TORC1 signaling, which is a central regulator of cell
Purinergic signaling in the pulmonary neuroepithelial body microenvironment unraveled by live cell imaging
De Proost I, et al.
Faseb Journal, 23(4), 1153-1160 (2009)
Mechanically induced chromatin condensation requires cellular contractility in mesenchymal stem cells
Heo SJ, et al.
Biophysical Journal, 111(4), 864-874 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service