Skip to Content
Merck
All Photos(1)

Key Documents

27840

Sigma-Aldrich

Corticosterone

≥98.5% (HPLC)

Synonym(s):

11β,21-Dihydroxy-4-pregnene-3,20-dione, 11β,21-Dihydroxyprogesterone, 4-Pregnene-11β,21-diol-3,20-dione, Kendall’s Compound B, Reichstein’s Substance H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
Beilstein:
2339601
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥98.5% (HPLC)

form

powder or crystals

optical activity

[α]20/D +223±3°, c = 1% in ethanol

mp

179-183 °C (lit.)
179-183 °C

functional group

ketone

SMILES string

C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO

InChI

1S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,14-17,19,22,24H,3-8,10-11H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1

InChI key

OMFXVFTZEKFJBZ-HJTSIMOOSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Corticosterone (Cort) is a glucocorticoid hormone. It is also the principal hormone that mediates stress responses in birds. It is the primary adrenal cortical steroid in rat plasma. The influence of maternal Cort exposure on the developing placenta has been studied in a mouse model.

Application

Corticosterone was used to study the adrenoceptor signaling in rats, lizards or in vitro. It was also used in immune suppression experiment.

Biochem/physiol Actions

Corticosterone is synthesized from cholesterol in the adrenal cortex that activates both mineralocorticoid and glucocorticoid receptors. The role of corticosterones is vital for reproductive processes including ovulation, lutenization, development of oocytes, parturition and lactation. The metabolism of corticosterone is modulated by the enzyme 11β-hydroxysteroid dehydrogenase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Corticosterone is soluble in chloroform and ethanol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corticosterone and brood abandonment in a passerine bird.
Ouyang JQ, et al.
Animal behaviour, 84(1), 261-268 (2012)
Stress, corticosterone responses and avian personalities.
Cockrem JF.
Journal of Ornithology, 148(2), 169-178 (2007)
Amanda J Rickard et al.
Journal of molecular endocrinology, 42(6), 449-459 (2009-01-23)
The mineralocorticoid receptor (MR) and glucocorticoid receptor are ligand-activated transcription factors that have important physiological and pathophysiological actions in a broad range of cell types including monocytes and macrophages. While the glucocorticoids cortisol and corticosterone have well-described anti-inflammatory actions on
A M Iacopino et al.
The Journal of biological chemistry, 265(18), 10177-10180 (1990-06-25)
Corticosterone was administered to normal and bilaterally adrenalectomized rats (250-300 g), and hormonal regulation of brain calbindin-D28k (CaBP28k) levels was investigated by radioimmunoassay for CaBP28k protein and by slot and Northern blot analyses for CaBP28k mRNA. The specificity of the
D F DeNardo et al.
Hormones and behavior, 27(2), 184-199 (1993-06-01)
Adrenal corticoid release is a major component of the stress response which can affect many body functions including behavior. The purpose of our studies was to examine the effects of corticosterone (B) on both the agonistic and courtship components of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service