P2613
ProteoMass™ P14R MALDI-MS Standard
vial of 10 nmol
Synonym(s):
P14R
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
Quality Level
mol wt
monoisotopic mol wt 1,532.8582 Da
packaging
vial of 10 nmol
storage temp.
−20°C
Amino Acid Sequence
Pro14-Arg
Application
Peptide calibration standard for MALDI-MS instruments.
Legal Information
ProteoMass is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Clinica chimica acta; international journal of clinical chemistry, 489, 154-161 (2018-12-12)
Chronic kidney disease (CKD) is a worldwide public health problem. To detect discriminating salivary peptide profiles between CKD patients undergoing haemodialysis (HD) and healthy controls (HCs) and to screen candidate biomarkers for CKD, we preliminarily explored the diagnostic potentiality of
Journal of dentistry, 61, 21-27 (2017-04-26)
To investigate differentially expressed salivary peptides in the development of early childhood caries (ECC) in 3-4 year-old children. Eighty-two caries-free children at baseline were followed-up for 1year, during which period 15 of them had developed ECC (Group C), whilst another
Annals of translational medicine, 8(11), 686-686 (2020-07-04)
Clinical and research interest in salivary peptidome and microbiota is ever-growing owing to its great value for diagnosis, risk assessment and prediction of prognosis in oral and systemic diseases. Saliva can be stimulated for the purpose of rapid collection, but
PloS one, 12(8), e0182712-e0182712 (2017-08-16)
This study aimed to identify differences of peptide profiles in stimulated whole saliva among children with and without occurrence of new carious lesions, and to provide a simple way for early diagnosis and prevention of the relapse of severe early
Novel serum peptide model revealed by MALDI-TOF-MS and its diagnostic value in early bladder cancer.
The International journal of biological markers, 35(3), 59-66 (2020-07-24)
Bladder cancer is the ninth most common cancer worldwide and has high morbidity and mortality. We aimed to search for potential serum peptide biomarkers and establish a diagnostic model for early bladder cancer. A total of 67 bladder cancer patients
Articles
One of the most important aspects of our ultra-pure MALDI matrix substances is their ability to dissolve rapidly and completely; a brief vortex mixing is typically sufficient.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service