Skip to Content
Merck
All Photos(1)

Key Documents

A0673

Sigma-Aldrich

Adrenocorticotropic Hormone Fragment 18-39 human

≥97% (HPLC)

Synonym(s):

CLIP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H165N27O36
CAS Number:
Molecular Weight:
2465.67
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C1CCCN1C(=O)C(Cc2ccccc2)NC(=O)C(C)NC(=O)C(CCC(O)=O)NC(=O)C(C)NC(=O)C(CO)NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(C)NC(=O)CNC(=O)C(CC(N)=O)NC(=O)C3CCCN3C(=O)C(Cc4ccc(O)cc4)NC(=O)C(NC(=O)C(CCCCN)NC(=O)C(NC(=O)C5CCCN5C(=O)C(N)CCCNC(N)=N)C(C)C)C(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(Cc6ccccc6)C(O)=O

InChI

1S/C112H165N27O36/c1-56(2)48-72(100(163)125-70(37-41-86(148)149)97(160)133-77(111(174)175)51-63-24-14-11-15-25-63)129-103(166)79-28-20-46-138(79)109(172)75(49-62-22-12-10-13-23-62)131-93(156)61(9)121-95(158)68(35-39-84(144)145)123-92(155)60(8)122-102(165)78(55-140)134-98(161)71(38-42-87(150)151)126-101(164)74(53-88(152)153)128-96(159)69(36-40-85(146)147)124-91(154)59(7)120-83(143)54-119-94(157)73(52-82(115)142)130-104(167)80-29-21-47-139(80)110(173)76(50-64-31-33-65(141)34-32-64)132-107(170)89(57(3)4)135-99(162)67(27-16-17-43-113)127-106(169)90(58(5)6)136-105(168)81-30-19-45-137(81)108(171)66(114)26-18-44-118-112(116)117/h10-15,22-25,31-34,56-61,66-81,89-90,140-141H,16-21,26-30,35-55,113-114H2,1-9H3,(H2,115,142)(H,119,157)(H,120,143)(H,121,158)(H,122,165)(H,123,155)(H,124,154)(H,125,163)(H,126,164)(H,127,169)(H,128,159)(H,129,166)(H,130,167)(H,131,156)(H,132,170)(H,133,160)(H,134,161)(H,135,162)(H,136,168)(H,144,145)(H,146,147)(H,148,149)(H,150,151)(H,152,153)(H,174,175)(H4,116,117,118)

InChI key

ZYDMZKPAPSZILB-UHFFFAOYSA-N

Gene Information

human ... POMC(5443)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe

General description

Adrenocorticotropic hormone (ACTH) is synthesized from its precursor pro-opiomelanocortin (POMC). It is released from anterior pituitary gland in response to corticotropin-releasing hormone (CRH) from hypothalamus. The gene encoding ACTH is localized on human chromosome 2p23.3.

Application

Adrenocorticotropic Hormone Fragment 18-39 human has been used for matrix-assisted laser desorption/ionization (MALDI)-TOF-MS and MALDI-MS.

Biochem/physiol Actions

Adrenocorticotropic hormone (ACTH) stimulates the synthesis and release of glucocorticoids and androgens through cAMP-dependent by a G-protein coupled receptor in adrenal gland. Overproduction of ACTH leads to marked skin pigmentation.
Facilitatory effect exerted on paradoxical sleep in the rat: stimulates insulin release.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long time storage (archiving) of peptide, protein and tryptic digest samples on disposable nano-coated polymer targets for MALDI MS
Stefan B, et al.
Annals of the Rheumatic Diseases, 8, 48-54 (2015)
Alex Steimle et al.
Journal of autoimmunity, 75, 82-95 (2016-08-04)
Cathepsin S (CTSS) is a lysosomal protease whose activity regulation is important for MHC-II signaling and subsequent activation of CD4+ T cell mediated immune responses. Dysregulation of its enzymatic activity or enhanced secretion into extracellular environments is associated with the
Patrick A Ott et al.
Nature, 547(7662), 217-221 (2017-07-06)
Effective anti-tumour immunity in humans has been associated with the presence of T cells directed at cancer neoantigens, a class of HLA-bound peptides that arise from tumour-specific mutations. They are highly immunogenic because they are not present in normal tissues
Transcriptomics and proteomics analyses of the PACAP38 influenced ischemic brain in permanent middle cerebral artery occlusion model mice
Motohide H, et al.
Journal of Neuroinflammation, 10(1), 791-791 (2013)
Patient Education: A Practical Approach
Doyle D
Annals of the Rheumatic Diseases, 52(10), 701-701 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service