Skip to Content
Merck
All Photos(1)

Key Documents

54793

Supelco

2-(4-Hydroxyphenylazo)benzoic acid

matrix substance for MALDI-MS, ≥99.5%

Synonym(s):

2-(4′-Hydroxybenzeneazo)benzoic acid, 4′-Hydroxyazobenzene-2-carboxylic acid, HABA, HBABA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4N=NC6H4CO2H
CAS Number:
Molecular Weight:
242.23
Beilstein:
1842696
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.5% (HPLC)
≥99.5%

analyte functional class(es)

polymers

analyte chemical class(es)

glycans, peptides, proteins

technique(s)

MALDI-MS: suitable

mp

204-208 °C (lit.)
204-208 °C

solubility

ethanol: 20 mg/mL, clear

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

≥2000 at 337 nm (mol. absorption)

SMILES string

Oc1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C13H10N2O3/c16-10-7-5-9(6-8-10)14-15-12-4-2-1-3-11(12)13(17)18/h1-8,16H,(H,17,18)/b15-14+

InChI key

DWQOTEPNRWVUDA-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(4-Hydroxyphenylazo)benzoic acid) (HABA), can be used for desorption of proteins, peptides and glycoproteins, thereby making it very advantageous for matrix-assisted laser desorption ionization mass spectrometry.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix-assisted laser desorption ionization mass spectrometry with 2-(4-hydroxyphenylazo) benzoic acid matrix.
Juhasz, Peter, Catherine E. Costello, and Klaus Biemann.
Journal of the American Society For Mass Spectrometry, 4 (5), 399-409 (1993)
K J Welham et al.
Rapid communications in mass spectrometry : RCM, 12(4), 176-180 (1998-03-11)
A variety of gram-positive and gram-negative intact bacterial cells have been analysed by matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) and shown to provide fingerprint mass spectra with discrete peaks being observed over the mass range from 3 to 40 kDa.
Joan-Antoni Farrera et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(7), 2277-2285 (2007-12-29)
The tautomeric equilibria of 2-(4'-hydroxyphenylazo)benzoic acid (HABA) and 2-(3',5'-dimethyl-4'-hydroxyphenylazo)benzoic acid (3',5'-dimethyl-HABA) have been studied by a combination of spectroscopic and computational methods. For neutral HABA in solvents of different polarity (toluene, chloroform, DMSO, DMF, butanol, and ethanol) the azo tautomer
M A Domin et al.
Rapid communications in mass spectrometry : RCM, 13(4), 222-226 (1999-03-31)
The ability to rapidly identify the taxonomic class of the wide variety of microorganisms involved in human and animal disease is becoming increasingly important, especially with the increasing development of resistance to the antibiotics which form the main defence against
Juha A E Määttä et al.
Chembiochem : a European journal of chemical biology, 9(7), 1124-1135 (2008-04-03)
Chicken avidin is a key component used in a wide variety of biotechnological applications. Here we present a circularly permuted avidin (cpAvd4-->3) that lacks the loop between beta-strands 3 and 4. Importantly, the deletion of the loop has a positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service