Skip to Content
Merck
All Photos(2)

Key Documents

78246

Supelco

Curcumin

matrix substance for MALDI-MS, ≥99.5% (HPLC)

Synonym(s):

(E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H3(OCH3)CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
Colour Index Number:
75300
Beilstein:
2306965
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS

Quality Level

vapor density

13 (vs air)

Assay

≥99.5% (HPLC)

form

powder

technique(s)

MALDI-MS: suitable

mp

183-188 °C

cation traces

Na: ≤50 mg/kg

storage temp.

2-8°C

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+

InChI key

VFLDPWHFBUODDF-FCXRPNKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Hybrid Ethylcellulose Polymeric Films: Ag(I)-Based Components and Curcumin as Reinforcing Ingredients for Enhanced Food Packaging Properties: This research discusses the development of hybrid ethylcellulose films integrating curcumin to improve food packaging, emphasizing the analytical chemistry aspects like ingredient compatibility and performance evaluation (Crispini A et al., 2024).

  • Intelligent double-layer film based on gellan gum/modified anthocyanin/curcumin/sodium alginate/zinc oxide for monitoring shrimp freshness: This article describes the creation of an intelligent film incorporating curcumin for real-time monitoring of shrimp freshness, showcasing a practical application of curcumin in food safety and shelf-life extension (Li C et al., 2024).

  • Cyclodextrin-grafted redox-responsive hydrogel mediated by disulfide bridges for regulated drug delivery: This study examines a curcumin-incorporated cyclodextrin hydrogel, highlighting the use of curcumin in developing advanced drug delivery systems with potential applications in targeted therapy (Xu X et al., 2024).

Biochem/physiol Actions

A natural phenolic compound responsible for the yellow color of turmeric. Potent anti-tumor agent that acts against highly diverse tumor-specific pathways. Anti-inflammatory and anti-oxidant. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cyclooxygenase (COX-2) and 5-lipoxygenase (5-LOX).
A natural phenolic compound. Potent anti-tumor agent having anti-inflammatory and anti-oxidant properties. Curcumin has been cited as a potential chemopreventive agent, in addition to its chemotherapeutic activity. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cycloxygenase and lipoxygenase. Easily penetrates into the cytoplasm of cells, accumulating in membranous structures such as plasma membrane, endoplasmic reticulum and nuclear envelope.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bharat B Aggarwal et al.
Molecular nutrition & food research, 57(9), 1529-1542 (2013-07-13)
Turmeric, a dried powder derived from the rhizome of Curcuma longa, has been used for centuries in certain parts of the world and has been linked to numerous biological activities including antioxidant, anti-inflammatory, anticancer, antigrowth, anti-arthritic, anti-atherosclerotic, antidepressant, anti-aging, antidiabetic
Shahar Lev-Ari et al.
The Journal of nutritional biochemistry, 25(8), 843-850 (2014-05-20)
Non-small cell lung cancer (NSCLC) is the leading cause of cancer-related mortality. Curcumin is involved in various biological pathways leading to inhibition of NSCLC growth. The purpose of this study was to evaluate the effect of curcumin on expression of
Anggakusuma et al.
Gut, 63(7), 1137-1149 (2013-08-02)
Hepatitis C virus (HCV) infection causes severe liver disease and affects more than 160 million individuals worldwide. People undergoing liver organ transplantation face universal re-infection of the graft. Therefore, affordable antiviral strategies targeting the early stages of infection are urgently
Vivian Soetikno et al.
Drug discovery today, 18(15-16), 756-763 (2013-05-09)
Diabetic nephropathy is characterized by a plethora of signaling abnormalities. Recent trials have suggested that intensive glucose-lowering treatment leads to hypoglycemic events, which can be dangerous. Curcumin is the active ingredient of turmeric, which has been widely used in many
Yoshikane Yamauchi et al.
Phytotherapy research : PTR, 28(5), 728-735 (2013-08-15)
Resistance to erlotinib in lung cancer cases includes T790M mutant epidermal growth factor receptor and c-Met gene amplification, but other unknown mechanisms account for about 30% of the resistance. Activation of the nuclear factor kappa B (NFkappaB)-related pathways in association

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service