Skip to Content
Merck
All Photos(1)

Key Documents

46278

Supelco

trans-Ferulic acid

matrix substance for MALDI-MS, ≥99.0% (HPLC)

Synonym(s):

trans-4-Hydroxy-3-methoxycinnamic acid, Ferulic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH=CHCO2H
CAS Number:
Molecular Weight:
194.18
Beilstein:
1570363
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (HPLC)

form

powder

analyte chemical class(es)

peptides, proteins

technique(s)

MALDI-MS: suitable

color

slightly yellow

mp

168-172 °C (lit.)
171-175 °C

cation traces

Ba: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

≥2000 at 337 nm (mol. absorption)

SMILES string

OC(/C=C/C1=CC(OC)=C(O)C=C1)=O

InChI

1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+

InChI key

KSEBMYQBYZTDHS-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hydroxycinnamic acid found in plant-based foods. Antioxidant.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengfeng Wu et al.
Critical reviews in food science and nutrition, 53(5), 451-463 (2013-02-09)
Brown rice, unmilled or partly milled, contains more nutritional components than ordinary white rice. Despite its elevated content of bioactive components, brown rice is rarely consumed as a staple food for its dark appearance and hard texture. The germination of
Sonia Trombino et al.
Journal of agricultural and food chemistry, 52(8), 2411-2420 (2004-04-15)
Although an antioxidant mechanism has been involved in the beneficial effects of ferulic acid in human diseases, there are few reports on the antioxidant properties of this compound in isolated membranes and intact cells. Here, we evaluated the ability of
Ping Geng et al.
Journal of separation science, 40(4), 991-998 (2016-12-13)
Sugarcane rind contains some functional phenolic acids. The separation of these compounds from sugarcane rind is able to realize the integrated utilization of the crop and reduce environment pollution. In this paper, a novel protocol based on interfacing online solid-phase
J P Rosazza et al.
Journal of industrial microbiology, 15(6), 457-471 (1995-12-01)
In this review we examine the fascinating array of microbial and enzymatic transformations of ferulic acid. Ferulic acid is an extremely abundant, preformed phenolic aromatic chemical found widely in nature. Ferulic acid is viewed as a commodity scale, renewable chemical
Helle Harder et al.
European journal of nutrition, 43(4), 230-236 (2004-08-17)
Rye bread contributes an important part of the whole grain intake in the Scandinavian diet. Ferulic acid is the major phenolic compound in rye bran and is an antioxidant in vitro and may, therefore, contribute to cardioprotective effects of whole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service