Skip to Content
Merck
All Photos(2)

Key Documents

M3445

Sigma-Aldrich

Magnolol

≥90% (HPLC), from plant

Synonym(s):

2,2′-Bichavicol, 5,5′-Diallyl-2,2′-biphenyldiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Quality Level

Assay

≥90% (HPLC)

solubility

ethanol: soluble 1 mg/mL
methanol: soluble 1.5 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

Oc1ccc(CC=C)cc1-c2cc(CC=C)ccc2O

InChI

1S/C18H18O2/c1-3-5-13-7-9-17(19)15(11-13)16-12-14(6-4-2)8-10-18(16)20/h3-4,7-12,19-20H,1-2,5-6H2

InChI key

VVOAZFWZEDHOOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnolol is a bioactive neolignan extracted from the root and stem bark of different Chinese herbal Magnolia trees. It has a hydroxylated bi-phenoid structure like that of its structural isomer honokiol. It is a lipophilic compound.

Application

Magnolol has been used:
  • to test its effect on muscle atrophy and macrophage subtypes in sarcopenia mouse model
  • to test its inhibitory effect on human umbilical vein endothelial cells (HUVECs) in angiogenesis assay
  • to test its anti-fungal effects on growth, ultrastructure, and biofilm formation of Candida spp. strains
  • in the preparation of immobilized amino-functionalized mesoporous silica particles for prolonged-release studies

Biochem/physiol Actions

Magnolol exerts anti-depressant and, neurodegenerative properties. It is also considered to be a potent anti-cancer agent for its involvement in reducing cell proliferation, cell cytotoxicity, and inducing apoptosis.
Magnolol is a bioactive plant component isolated from the root and stem bark of Magnolia officinalis with antifungal, antibacterial and antioxidant effects. Magnolol also demonstrates anti-inflammatory activity by interferring with NF-κB signaling. It also appears to have anti-inflammatory properties related to vascular disorders such as atherosclerosis due to its ability to inhibit IL-6-induced STAT3 activation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanna Kuk et al.
Scientific reports, 7(1), 17820-17820 (2017-12-21)
Due to gravity the venous vasculature in the lower extremities is exposed to elevated pressure levels which may be amplified by obesity or pregnancy. As a consequence, venules dilate and may be slowly transformed into varicose or spider veins. In
Jawad Behbehani et al.
Microbial pathogenesis, 113, 209-217 (2017-10-28)
The incidence of oral candidosis has increased in recent years due to the escalation in HIV-infection, cancer treatments, organ transplantation, and diabetes. In addition, corticosteroid use, dentures, and broad-spectrum antibiotic use have also contributed to the problem. Treatment of oral
Development of a prolonged-release drug delivery system with magnolol loaded in amino-functionalized mesoporous silica
Stefanache A, et al.,
Applied Sciences, 237-237 (2017)
Yongtao Bai et al.
Steroids, 135, 73-78 (2018-03-21)
Evidence showed that the stress hormone corticosterone (CORT) injection resulted in dysregulation of the hypothalamic-pituitary-adrenal (HPA) axis implicated in major depressive disorder. Magnolol, main constituent identified in the barks of Magnolia officinalis, exerted antidepressant effects in a rat model of
Jianhong Zhang et al.
BioMed research international, 2019, 1847130-1847130 (2019-06-27)
Over years, various biological constituents are isolated from Traditional Chinese Medicine and confirmed to show multifunctional activities. Magnolol, a hydroxylated biphenyl natural compound isolated from Magnolia officinalis, has been extensively documented and shows a range of biological activities. Many signaling

Articles

Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer.

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service