Skip to Content
Merck
All Photos(2)

Key Documents

42612

Supelco

Honokiol

analytical standard

Synonym(s):

5,3′-Diallyl-2,4′-dihydroxybiphenyl, NSC 293100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1CC=C)-c2cc(CC=C)ccc2O

InChI

1S/C18H18O2/c1-3-5-13-7-9-18(20)16(11-13)14-8-10-17(19)15(12-14)6-4-2/h3-4,7-12,19-20H,1-2,5-6H2

InChI key

FVYXIJYOAGAUQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Honokiol is a natural biphenyl neolignan from magnolia extract. It is antiangiogenic, antitumor, and anxiolytic.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankit Kumar et al.
Future medicinal chemistry, 5(7), 809-829 (2013-05-09)
Honokiol (3,5-di-(2-propenyl)-1,1-biphenyl-2,2-diol) is a natural bioactive neolignan isolated from the genus Magnolia. In recent studies, honokiol has been observed to have anti-angiogenic, anticancer, anti-inflammatory, neuroprotective and GABA-modulating properties in vitro and in preclinical models. Honokiol and its analogs target multiple
Haiquan Wang et al.
Neuropharmacology, 86, 9-21 (2014-06-29)
Recently, increasing evidence has shown that cell cycle activation is a key factor of neuronal death and neurological dysfunction after traumatic brain injury (TBI). This study aims to investigate the effects of Honokiol, a cell cycle inhibitor, on attenuating the
Eun-Ryeong Hahm et al.
The Prostate, 74(4), 408-420 (2013-12-18)
We have shown previously that honokiol (HNK), a bioactive component of the medicinal plant Magnolia officinalis, inhibits growth of human prostate cancer cells in vitro and in vivo. However, the effect of HNK on androgen receptor (AR) signaling has not
Ying Ju Chen et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(4), 573-580 (2014-01-01)
Proinflammatory cytokine interleukin-1β (IL-1β) stimulates several mediators of cartilage degradation and plays an important role in the pathogenesis of osteoarthritis (OA). Honokiol, a low molecular weight natural product isolated from the Magnolia officinalis, has been shown to possess anti-inflammatory effect.
Levi E Fried et al.
Antioxidants & redox signaling, 11(5), 1139-1148 (2009-02-11)
Honokiol is a small-molecule polyphenol isolated from the genus Magnolia. It is accompanied by other related polyphenols, including magnolol, with which it shares certain biologic properties. Recently, honokiol has been found to have antiangiogenic, antiinflammatory, and antitumor properties in preclinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service