Skip to Content
Merck
All Photos(4)

Key Documents

C1386

Sigma-Aldrich

Curcumin

from Curcuma longa (Turmeric), powder

Synonym(s):

(E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H3(OCH3)CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
Colour Index Number:
75300
Beilstein:
2306965
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

Curcuma longa (Turmeric)

Quality Level

vapor density

13 (vs air)

form

powder

concentration

≥65% (HPLC)

mp

175 °C

solubility

ethanol: 10 mg/mL
DMSO: >11 mg/mL (lit.)(lit.)
0.5 M NaOH: soluble (then immediately dilute in PBS [lit.])(lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+

InChI key

VFLDPWHFBUODDF-FCXRPNKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Curcumin is a yellow-colored polyphenol obtained from the rhizome of Curcuma longa L. It constitutes an important part of various diets and herbal medicines, especially in the Asian continent for its various salubrious effects on human health.[1][2][3]

Application

Curcumin has been used:
  • to study its effect on the consequences and mechanism involved in the suppression of human homeobox gene NKX3.1 in the prostate cancer cell LNCaP[1]
  • to examine its effect on stress in pigs by having an inhibitory impact on the serum cortisol concentration, hippocampal nitric oxide production, and brain-derived neurotrophic factor expression[2]
  • to study its effect as a dietary supplement on the growth, immunity, antioxidant activity, and disease resistance in Oreochromis niloticus[3]
  • to analyze its protective effect on the organotypic hippocampal slice cultures against the synaptic toxicity caused by amyloid beta peptides (Aβ1–42)[4]
  • to examine the possibility of its non-toxic concentrations to decrease the inflammation caused by interleukin-1beta (IL-1β) in cartilage explant cultures[5]
  • to study its protective impact against intestinal ischemia-reperfusion injury in rats[6]
  • to measure its antibacterial activity in vitro[3]
  • to determine its preventative effects for Alzheimer′s disease (AD) in mice[7]
  • as a bifunctional agent for generation and validation of the erythroid 2-related factor 2 (Nrf2) reporter system[8]
  • to study its in vitro inhibitory effects on human liver glucuronidation activity[9]
  • to evaluate its effects on Parkinson′s disease (PD)-like phenotypes[10]

Biochem/physiol Actions

A natural phenolic compound responsible for the yellow color of turmeric. Potent anti-tumor agent that acts against highly diverse tumor-specific pathways. Anti-inflammatory and anti-oxidant. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cyclooxygenase (COX-2) and 5-lipoxygenase (5-LOX).
A natural phenolic compound. Potent anti-tumor agent having anti-inflammatory and anti-oxidant properties. Curcumin has been cited as a potential chemopreventive agent, in addition to its chemotherapeutic activity. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cycloxygenase and lipoxygenase. Easily penetrates into the cytoplasm of cells, accumulating in membranous structures such as plasma membrane, endoplasmic reticulum and nuclear envelope.
Curcumin possesses anti-inflammatory, cancer prophylactic, and anti-oxidant properties. It also exhibits antagonistic nature towards the cluster of colon, breast, and blood marrow cancer. It does so by means of having an inhibitory effect on the various associated enzymes such as cycloxygenase-2 (COX-2), lipoxygenase and ornithine decarboxylase.[1] It is also postulated to have anti-depressant, anti-stress and neuroprotective effects on humans and other animals.[2] It is being proclaimed to be used as a treatment for Alzheimer′s disease due to its ability to cause synaptic toxicity.[4] Additionally, it also shows anti-fungal, antiviral, anti-microbial, chemosensitizing, radiosensitizing, and wound healing activities.[6]

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of diets containing curcumin, epigallocatechin-3-gallate, docosahexaenoic acid and alpha-lipoic acid on amyloid load and inflammation in a male transgenic mouse model of Alzheimer's disease: Are combinations more effective?
Sharman MJ, et al.
Neurobiology of Disease, 124, 505-519 (2019)
In vitro inhibitory effects of Andrographis paniculata, Gynura procumbens, Ficus deltoidea, and Curcuma xanthorrhiza extracts and constituents on human liver glucuronidation activity
Husni Z, et al.
Pharmacognosy magazine, 13(Suppl 2), S236-S236 (2017)
In vivo imaging of early signs of dopaminergic neuronal death in an animal model of Parkinson's disease
Rizzi N, et al.
Neurobiology of Disease, 114, 74-84 (2018)
Curcumin effectively rescued Parkinson?s disease-like phenotypes in a novel drosophila melanogaster model with dUCH Knockdown
Nguyen TT, et al.
Oxidative Medicine and Cellular Longevity, 2018 (2018)
Curcumin's biphasic hormetic response on proteasome activity and heat-shock protein synthesis in human keratinocytes
Ali RE and Rattan SIS
Annals of the New York Academy of Sciences, 1067(1), 394-399 (2006)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Can ı use this Curcumin for loading in the liposome?

    1 answer
    1. The research paper "Encapsulation of Curcumin-Loaded Liposomes for Colonic Drug Delivery in a pH-Responsive Polymer Cluster Using a pH-Driven and Organic Solvent-Free Process" (De Leo, V., et. al.) cites the use of Curcumin "purchased from Sigma-Aldrich".

      Helpful?

  4. what is % purity of the product

    1 answer
    1. The purity will be listed on the lot-specific Certificate of Analysis. Please navigate to the ‘DOCUMENTATION’ section of the Product Detail Page to access a Certificate:
      https://www.sigmaaldrich.com/product/sigma/c1386#product-documentation

      Helpful?

  5. is this curcumin nanoparticle?

    1 answer
    1. The product is specified to be a 'powder' but the particle size is not characterized. It is likely more on the order of microns than nanometers, but it can't be guaranteed either way.

      Helpful?

  6. What are the recommended methods for dissolving curcumin powder (C1386)?

    1 answer
    1. Dissolving curcumin (product C1386) can be challenging. Below are some recommendations for preparing standard solutions:
      • Ethanol: 10mg/mL
      • DMSO: >11 mg/mL (according to available literature)
      • 0.5 M NaOH: Soluble (then immediately dilute in PBS according to literature)
      Additionally, here are two methods that might be helpful:
      Method 1: Use DMSO to create a stock solution at 1000 times the working concentration of curcumin.

      Helpful?

  7. Hello, I am currently looking for curcumin powder that I can use as a standard for UV spectrophotometer quantification. What is the difference between the product that is offered here compared to 08511-10 MG Curcumin product?

    1 answer
    1. Both items are research grade products. Product 08511 is assigned a minimum purity specification of 98% and is tested for use as an an analytical standard in HPLC and GC. The C1386 is assigned a minimum purity specification of 65% and is not tested for any specific application. The major differences will lie in the purity and testing for each product. Please see the sample Certificate of Analysis for both items:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/C1386-10G0000176612.pdf
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/08511-BULKWXBD9237V.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service