Skip to Content
Merck
All Photos(1)

Key Documents

C0567

Sigma-Aldrich

(−)-Catechin

from green tea, ≥97% (HPLC), powder, oxidative stress inhibitor

Synonym(s):

(−)-trans-3,3′,4′,5,7-Pentahydroxyflavane, (2S,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

(−)-Catechin, ≥97% (HPLC), from green tea

biological source

green tea

Assay

≥97% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m1/s1

InChI key

PFTAWBLQPZVEMU-HIFRSBDPSA-N

Gene Information

human ... BACE1(23621)

Looking for similar products? Visit Product Comparison Guide

General description

Catechins are major flavonoids present in green tea accounting to 90%. (−)-catechin is comparatively less bioavailable than the (+)-catechin for intestinal absorption.

Application

(-)-Catechin has been used:
  • to test its binding affinity towards estrogen receptor
  • as a reference standard in high-performance liquid chromatography for quantification of polyphenols from plant extracts (HPLC)
  • as a reference standard in kinetic studies to quantify antioxidants from Iranian green tea leaves

Biochem/physiol Actions

Catechins have wide biological functionality. They induce antiviral and anticancer activities. Catechins also play a key role in regulating carbohydrate and lipid metabolism. It prevents fat accumulation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogen-like effects of wine extracts on nitric oxide synthesis in human endothelial cells
Simoncini T, et al.
Maturitas, 70(2), 169-175 (2011)
Rajapandiyan Krishnamoorthy et al.
Biomolecules, 9(12) (2019-12-11)
Human gut microbes are a profitable tool for the modification of food compounds into biologically active metabolites. The biological properties of catechins have been extensively investigated. However, the bioavailability of catechin in human blood plasma is very low. This study
Green tea catechins: Their use in treating and preventing infectious diseases
Reygaert WC
BioMed Research International, 2018 (2018)
Veronica Sanda Chedea et al.
Journal of the science of food and agriculture, 97(8), 2516-2523 (2016-11-02)
The effect of a diet containing 15% grape pomace (GP) on the general health status and milk quality of dairy cows was assessed by plasma biochemistry and total polyphenol (TP) content, milk polyphenols, milk composition and milk protein fractions. Among
Ingestion of a tea rich in catechins leads to a reduction in body fat and malondialdehyde-modified LDL in men
Nagao T, et al.
American Journal of Clinical Nutrition, 81(1), 122-129 (2005)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service